Skip to Content
Merck

Skip To

8.22285

Sodium nitrite

cryst. for synthesis

Synonym(s):

Sodium nitrite

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Pack SizeSKUAvailabilityPrice

About This Item

Empirical Formula (Hill Notation):
NNaO2
CAS Number:
Molecular Weight:
69.00
UNSPSC Code:
12352302
EC Index Number:
231-555-9
NACRES:
NA.22
MDL number:
Grade:
synthesis grade
Solubility:
820 g/L
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


grade

synthesis grade

Quality Level

potency

85 mg/kg LD50, oral (Rat)

pH

9 (20 °C, 100 g/L in H2O)

mp

280 °C (decomposition)

solubility

820 g/L

density

2.1 g/cm3 at 20 °C

bulk density

1200 kg/m3

storage temp.

2-30°C

SMILES string

[Na+].N(=O)[O-]

InChI

1S/HNO2.Na/c2-1-3;/h(H,2,3);/q;+1/p-1

InChI key

LPXPTNMVRIOKMN-UHFFFAOYSA-M


Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
31443799416237213
solubility

820 g/L

solubility

-

solubility

-

solubility

-

Quality Level

200

Quality Level

200

Quality Level

100

Quality Level

200

grade

synthesis grade

grade

ACS reagent, puriss. p.a.

grade

ACS reagent, anhydrous

grade

ACS reagent

mp

280 °C (decomposition)

mp

271 °C (lit.)

mp

271 °C (lit.)

mp

271 °C (lit.)

pH

9 (20 °C, 100 g/L in H2O)

pH

9

pH

9

pH

-

density

2.1 g/cm3 at 20 °C

density

-

density

-

density

-


Still not finding the right product?

Explore all of our products under Sodium nitrite


signalword

Danger

Hazard Classifications

Acute Tox. 3 Oral - Aquatic Acute 1 - Eye Irrit. 2 - Ox. Sol. 3

Storage Class

5.1B - Oxidizing hazardous materials

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable



Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Global Trade Item Number

SKUGTIN
822285010004022536487724
822285905004022536738109
822285000004022536487717
822285500004022536487748
822285100004022536487731