Skip to Content
Merck
All Photos(1)

Documents

31443

Sigma-Aldrich

Sodium nitrite

puriss. p.a., ACS reagent, reag. Ph. Eur., ≥99%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
NaNO2
CAS Number:
Molecular Weight:
69.00
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:

grade

ACS reagent
puriss. p.a.

Quality Level

Agency

reag. Ph. Eur.

vapor pressure

<0.0001 hPa ( 25 °C)

Assay

≥99%

form

powder or crystals

autoignition temp.

914 °F

impurities

≤0.001% heavy metals (as Pb)

pH

9

mp

271 °C (lit.)

anion traces

chloride (Cl-): ≤50 mg/kg
sulfate (SO42-): ≤50 mg/kg

cation traces

Ca: ≤20 mg/kg
Fe: ≤10 mg/kg
K: ≤50 mg/kg

SMILES string

[Na+].[O-]N=O

InChI

1S/HNO2.Na/c2-1-3;/h(H,2,3);/q;+1/p-1

InChI key

LPXPTNMVRIOKMN-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

General description

Combined effect of NaNO2 and NaCl on the growth of Listeria monocytogenes in tryptone soya broth has been investigated using automated turbidimetric system. Its carcinogenic effects in F-344 rats have been evaluated.

Application

Sodium nitrite may be used as a standard for the quantitative determination of nitrite in culture medium. It may be used in the preparation of isopropyl nitrite, via reaction with isoamyl alcohol.

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Oral - Aquatic Acute 1 - Eye Irrit. 2 - Ox. Sol. 3

Storage Class Code

5.1B - Oxidizing hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Alyssa Gill et al.
Analytica chimica acta, 1077, 167-173 (2019-07-17)
Nitrite is a naturally occurring species present in various food samples and also present in our bodies as a product of nitric oxide (NO) oxidation. Considering the ubiquity of nitrite, its determination is of great importance in both biological and
Noga Gal et al.
The journal of physical chemistry. B, 122(22), 5820-5834 (2018-05-05)
Core-shell nanoparticles receive much attention for their current and potential applications in life sciences. Commonly, a dense shell of hydrated polymer, a polymer brush, is grafted to improve colloidal stability of functional nanoparticles and to prevent protein adsorption, aggregation, cell
A Maekawa et al.
Food and chemical toxicology : an international journal published for the British Industrial Biological Research Association, 20(1), 25-33 (1982-02-01)
The carcinogenicity of sodium and of sodium nitrate was examined in F-344 rats. Sodium nitrite was administered in the drinking-water for 2 yr at levels of 0.125 or 0.25%. Sodium nitrate was given in the diet at levels 2.5 or
Hsin-Tzu Tai et al.
Polymers, 11(8) (2019-08-21)
We investigated the self-assembled structures and photoresponsive and crystallization behaviors of supramolecules composed of 4-methoxy-4'-hydroxyazobenzene (Azo) molecules and polyethylene glycol (PEG) that were formed through hydrogen-bonding interactions. The Azo/PEG complexes exhibited the characteristics of photoresponse and crystallization, which originated from
Fei Yang et al.
Journal of hazardous materials, 406, 124774-124774 (2020-12-15)
Futility of traditional advanced oxidation processes (AOPs) in saline wastewater treatment has stimulated the quest for novel "halotolerant" chemical oxidation technology. Acetylacetone (AA) has proven to be a potent photo-activator in the degradation of dyes, but the applicability of UV/AA

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service