Skip to Content
Merck

Skip To

65035-M

Trypsin Inhibitor, Soybean

Synonym(s):

Trypsin Inhibitor, Soybean

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Pack SizeSKUAvailabilityPrice

About This Item

CAS Number:
MDL number:
NACRES:
NA.77
UNSPSC Code:
12352202
Form:
lyophilized solid
Quality level:
Storage condition:
OK to freeze, desiccated
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


form

lyophilized solid

Quality Level

specific activity

≥4000 units/mg protein

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze, desiccated

color

off-white

solubility

67 mM potassium phosphate buffer, pH 7.6: 5 mg/mL

storage temp.

−20°C

General description

A reversible inhibitor of trypsin that binds to trypsin in a stoichiometric, pH-dependent manner (Kd = 500 nM at pH 7.8).
Reversible inhibitor of trypsin that binds to trypsin in a stoichiometric, reversible and pH-dependent manner (Kd = 0.5 nM at pH 7.8). Also inhibits, to a lesser degree, chymotrypsin, kallikrein, plasmin, and other serine proteases.

Biochem/physiol Actions

Kd = 500 nM at pH 7.8 against trypsin
Primary Target
trypsin

Preparation Note

Following reconstitution, refrigerate (4°C). Stock solutions are stable for up to 1 week at 4°C.

Other Notes

Lilly, C.M., et al. 1993. J. Clin. Invest.91, 235.
Uchino, T., et al. 1993. J. Biol. Chem.268, 527.
Birk, Y. 1976. Methods Enyzmol.45, 700.
One unit is defined as the amount of protein that will inhibit 1 unit of trypsin activity using BAEE as a substrate at 25°C, pH 7.6.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Toxicity: Standard Handling (A)

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
T6522T900393620
form

lyophilized solid

form

powder

form

lyophilized powder

form

lyophilized powder

Quality Level

100

Quality Level

300

Quality Level

300

Quality Level

100

manufacturer/tradename

Calbiochem®

manufacturer/tradename

-

manufacturer/tradename

-

manufacturer/tradename

-

storage temp.

−20°C

storage temp.

2-8°C

storage temp.

2-8°C

storage temp.

−20°C

solubility

67 mM potassium phosphate buffer, pH 7.6: 5 mg/mL

solubility

balanced salt solution: 1 mg/mL, serum-free medium: soluble

solubility

H2O: >10 mg/mL, phosphate buffer: >10 mg/mL (hazy above 10 mg/ml), soluble

solubility

0.1 M phosphate pH 7.6: 5 mg/mL, clear, colorless to almost colorless

storage condition

OK to freeze, desiccated

storage condition

-

storage condition

-

storage condition

-


pictograms

Health hazard

signalword

Danger

hcodes

Hazard Classifications

Resp. Sens. 1 - Skin Sens. 1

Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable



Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Global Trade Item Number

SKUGTIN
65035-100MG07790788052164
65035-1GM04055977184501

Questions

Reviews

No rating value

Active Filters