Select a Size
| Size/SKU | Availability | Price |
|---|
About This Item
Quality Segment
form
liquid
feature
Snap-N-Spike®/Snap-N-Shoot®
packaging
ampule of 1 mL
manufacturer/tradename
Cerilliant®
drug control
Narcotic Licence Schedule D (Switzerland); psicótropo (Spain); Decreto Lei 15/93: Tabela IIB (Portugal)
concentration
100 μg/mL in heptane
technique(s)
gas chromatography (GC): suitable, liquid chromatography (LC): suitable
application(s)
cannabis testing
format
single component solution
storage temp.
−20°C
SMILES string
O1C(C2C(c3c1cc(cc3O)CCCCC)C=C(CC2)C)(C)C
InChI
1S/C21H30O2/c1-5-6-7-8-15-12-18(22)20-16-11-14(2)9-10-17(16)21(3,4)23-19(20)13-15/h11-13,16-17,22H,5-10H2,1-4H3
InChI key
CYQFCXCEBYINGO-UHFFFAOYSA-N
General description
Other Notes
Legal Information
1 of 1
This Item | |||
|---|---|---|---|
| technique(s) gas chromatography (GC): suitable, liquid chromatography (LC): suitable | technique(s) gas chromatography (GC): suitable, liquid chromatography (LC): suitable | technique(s) gas chromatography (GC): suitable, liquid chromatography (LC): suitable | technique(s) gas chromatography (GC): suitable, liquid chromatography (LC): suitable |
| form liquid | form liquid | form liquid | form liquid |
| format single component solution | format single component solution | format single component solution | format single component solution |
| Quality Level 100 | Quality Level 300 | Quality Level 300 | Quality Level 300 |
| storage temp. −20°C | storage temp. −20°C | storage temp. −20°C | storage temp. −20°C |
| manufacturer/tradename Cerilliant® | manufacturer/tradename Cerilliant® | manufacturer/tradename Cerilliant® | manufacturer/tradename Cerilliant® |
signalword
Danger
hcodes
Hazard Classifications
Aquatic Acute 1 - Aquatic Chronic 1 - Asp. Tox. 1 - Flam. Liq. 2 - Skin Irrit. 2 - STOT SE 3
target_organs
Central nervous system
Storage Class
3 - Flammable liquids
wgk
WGK 2
flash_point_f
24.8 °F - closed cup
flash_point_c
-4 °C - closed cup
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.






