Select a Size
| Size/SKU | Availability | Price |
|---|
About This Item
grade
certified reference material
Quality Segment
form
liquid
feature
(Snap-N-Spike®)
packaging
ampule of 1 mL
manufacturer/tradename
Cerilliant®
drug control
psicótropo (Spain); Decreto Lei 15/93: Tabela IIB (Portugal)
concentration
1.0 mg/mL in methanol
technique(s)
gas chromatography (GC): suitable, liquid chromatography (LC): suitable
application(s)
cannabis testing
forensics and toxicology
format
single component solution
storage temp.
−20°C
SMILES string
OC1=C2C3C(CCC(C(O)=O)=C3)C(C([2H])([2H])[2H])(C([2H])([2H])[2H])OC2=CC(CCCCC([2H])([2H])[2H])=C1
InChI
1S/C21H28O4/c1-4-5-6-7-13-10-17(22)19-15-12-14(20(23)24)8-9-16(15)21(2,3)25-18(19)11-13/h10-12,15-16,22H,4-9H2,1-3H3,(H,23,24)/i1D3,2D3,3D3
InChI key
YOVRGSHRZRJTLZ-GQALSZNTSA-N
General description
Legal Information
1 of 1
This Item | |||
|---|---|---|---|
| technique(s) gas chromatography (GC): suitable, liquid chromatography (LC): suitable | technique(s) gas chromatography (GC): suitable, liquid chromatography (LC): suitable | technique(s) gas chromatography (GC): suitable, liquid chromatography (LC): suitable | technique(s) gas chromatography (GC): suitable, liquid chromatography (LC): suitable |
| format single component solution | format single component solution | format single component solution | format single component solution |
| form liquid | form liquid | form liquid | form liquid |
| Quality Level 300 | Quality Level 300 | Quality Level 300 | Quality Level 300 |
| grade certified reference material | grade certified reference material | grade certified reference material | grade certified reference material |
| storage temp. −20°C | storage temp. -10 to -25°C | storage temp. 2-8°C | storage temp. 2-8°C |
Still not finding the right product?
Explore all of our products under (±)-11-nor-9-Carboxy-Δ9-THC-D9 solution
signalword
Danger
Hazard Classifications
Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Flam. Liq. 2 - STOT SE 1
target_organs
Eyes
Storage Class
3 - Flammable liquids
wgk
WGK 1
flash_point_f
49.5 °F
flash_point_c
9.7 °C
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.




