Skip to Content
Merck

Skip To

TP0010

FLAG® HA Tandem Affinity Purification Kit

Synonym(s):

Anti-ddddk, Anti-dykddddk

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Pack SizeSKUAvailabilityPrice

About This Item

NACRES:
NA.32
UNSPSC Code:
12352200
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


storage temp.

2-8°C

Quality Level

General description

TAP technology is a recent development in protein purification. This technology incorporates tandem-linked affinity tags into genes of interest so that high purity fusion proteins can be isolated through two consecutive affinity purification steps. The technology is particularly useful for the isolation and identification of protein complexes by adding a tandem affinity tag to a known "bait" protein to pull down endogenous proteins that interact with the targeted protein of interest.

Application

The FLAG® HA Tandem Affinity Purification Kit is designed for the isolation of high purity FLAG-HA dual-tagged fusion proteins from complex matrix, such as cell lysates and tissue homogenates. The kit is particularly suitable for isolation of protein complexes using TAP (Tandem Affinity Purification) technology.

Sutitable for mass spectrometry analysis and western blotting.

Learn more product details in our FLAG® application portal.

Legal Information

FLAG is a registered trademark of Merck KGaA, Darmstadt, Germany

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
FLAGIPT1F2426A2220
Quality Level

200

Quality Level

300

Quality Level

200

Quality Level

200

storage temp.

2-8°C

storage temp.

−20°C

storage temp.

−20°C

storage temp.

−20°C


Kit Components Also Available Separately

Product No.
Description
SDS & Pricing

  • EZview Red ANTI-FLAG® M2 Affinity Gel, clone M2
    SDS

  • Monoclonal Anti-HA−Agarose antibody produced in mouse, clone HA-7, purified immunoglobulin, PBS suspension
    SDS

  • RIPA Buffer
    SDS

  • 3X FLAG® Peptide, lyophilized powder
    SDS

  • Urea, 8 M (after reconstitution with 16 mL high purity water)
    SDS

Storage Class

10 - Combustible liquids

wgk

WGK 3

pictograms

Exclamation mark

signalword

Danger

Hazard Classifications

Aquatic Chronic 3 - ED ENV 1 - Eye Irrit. 2



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library


Related Content


Wei Li et al.
Cell, 140(4), 477-490 (2010-02-25)
Current models imply that the FERM domain protein Merlin, encoded by the tumor suppressor NF2, inhibits mitogenic signaling at or near the plasma membrane. Here, we show that the closed, growth-inhibitory form of Merlin accumulates in the nucleus, binds to
Junsong Zhang et al.
Journal of virology, 90(8), 3966-3980 (2016-02-05)
The viral ribonucleoprotein (vRNP) complex of influenza A viruses (IAVs) contains an RNA-dependent RNA polymerase complex (RdRp) and nucleoprotein (NP) and is the functional unit for viral RNA transcription and replication. The vRNP complex is an important determinant of virus
Karthik Ramachandran et al.
iScience, 25(1), 103722-103722 (2022-01-11)
SARS-CoV-2 is a newly identified coronavirus that causes the respiratory disease called coronavirus disease 2019 (COVID-19). With an urgent need for therapeutics, we lack a full understanding of the molecular basis of SARS-CoV-2-induced cellular damage and disease progression. Here, we



Global Trade Item Number

SKUGTIN
TP0010-5RXN04061837397981

Questions

Reviews

No rating value

Active Filters