Skip to Content
Merck

Skip To

FLAGIPT1

FLAG® Immunoprecipitation Kit

Synonym(s):

Anti-ddddk, Anti-dykddddk

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Pack SizeSKUAvailabilityPrice

About This Item

NACRES:
NA.32
UNSPSC Code:
12352200
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


usage

 kit sufficient for 50 reactions

Quality Segment

analyte chemical class(es)

proteins (FLAG® Affinity Gels)

technique(s)

immunoprecipitation (IP): suitable

shipped in

wet ice

storage temp.

−20°C

General description

Sigma′s FLAG Immunoprecipitation kit allows a rapid and efficient immunoprecipitation and elution of an active FLAG-tagged protein. The immunoprecipitation is performed with the ANTI-FLAG M2 affinity gel, which is a highly specific monoclonal antibody covalently attached to agarose resin. The use of affinity resin allows an efficient binding of FLAG-tagged proteins without the need for preliminary steps and calibrations. The immunoprecipitated FLAG-tagged proteins can be efficiently eluted from the resin with acidic conditions or by competition with FLAG peptide. The immunoprecipitated proteins can be detected for their size, post-translational modifications, and interactions on gel electrophoresis, and by activity assays.

Application

Learn more product details in our FLAG® application portal.

Features and Benefits

  • Utilizes highly specific ANTI-FLAG M2 affinity gel
  • No preliminary steps or calibrations
  • 3xFLAG peptide provides efficient and gentle elution through direct competition

Other Notes

All prodedures to be performed at 2-8°C.

Legal Information

3xFLAG is a trademark of Sigma-Aldrich Co. LLC
FLAG is a registered trademark of Merck KGaA, Darmstadt, Germany

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
F2426A2220F3290
technique(s)

immunoprecipitation (IP): suitable

technique(s)

affinity chromatography: suitable, immunoprecipitation (IP): suitable

technique(s)

affinity chromatography: suitable, immunoprecipitation (IP): suitable

technique(s)

-

usage

 kit sufficient for 50 reactions

usage

-

usage

-

usage

-

Quality Level

300

Quality Level

200

Quality Level

200

Quality Level

200

storage temp.

−20°C

storage temp.

−20°C

storage temp.

−20°C

storage temp.

2-8°C

shipped in

wet ice

shipped in

wet ice

shipped in

wet ice

shipped in

wet ice

analyte chemical class(es)

proteins (FLAG® Affinity Gels)

analyte chemical class(es)

proteins

analyte chemical class(es)

proteins

analyte chemical class(es)

-


Kit Components Also Available Separately

Product No.
Description
SDS & Pricing

  • ANTI-FLAG M2 affinity gel 1 mL
    SDS

  • 3X FLAG peptide 1 mg
    SDS

pictograms

Exclamation mark

Storage Class

10 - Combustible liquids

wgk

WGK 3

signalword

Danger

Hazard Classifications

Aquatic Chronic 3 - ED ENV 1 - Eye Irrit. 2



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library







Global Trade Item Number

SKUGTIN
FLAGIPT1-1KT04061833622902

Questions

Reviews

No rating value

Active Filters