Skip to Content
Merck
All Photos(1)

Key Documents

P1435

Sigma-Aldrich

Phosphatase, Acid from sweet potato

ammonium sulfate suspension, ≥10.0 units/mg protein (modified Warburg-Christian)

Synonym(s):

(Orthophosphoric-monoester phosphohydrolase [acid optimum] )

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
Enzyme Commission number:
EC Number:
MDL number:
UNSPSC Code:
12352204
NACRES:
NA.54

form

ammonium sulfate suspension

specific activity

≥10.0 units/mg protein (modified Warburg-Christian)

secondary activity

≤9.9 units/mg protein ATPase

mol wt

110 kDa

storage temp.

2-8°C

SMILES string

O(CC(O)C)CC#C

InChI

1S/C6H10O2/c1-3-4-8-5-6(2)7/h1,6-7H,4-5H2,2H3

InChI key

GZCWLCBFPRFLKL-UHFFFAOYSA-N

General description

Acid phosphatase from sweet potato is a phosphomonoesterase, which can appear in multiple molecular forms of similar molecular mass but with different isoelectric points.

Application

Potato acid phosphatase is used as a dephosphorylating reagent in thiochrome assays for the detection of thiamin in biological samples. In this assay, it was shown to be more effective than acid phosphatases from wheat germ or α-amylase.
Sweet potato acid phosphatase has been used in a study as a pre-column enzyme reactor via a covalent bond with glutaraldehyde and aminopropyl controlled-pore glass. It has also been used in a study to investigate the use of tyrosine, histidine, and cysteine as ligands for Mn(III) in sweet potato phosphatase.

Biochem/physiol Actions

Acid phosphatases (APase) are a family of enzymes that non-specifically catalyze the hydrolysis of monoesters and anhydrides of phosphoric acid to produce inorganic phosphate at an optimum pH of 4 to 7 by the following reaction: APaseR-PO4 + H2O --------------- R-OH + HOPO3 2+Their function in the production, transport, and recycling of phosphate is critical for the metabolic and energy transduction processes of the cell. As a group, Apases may be as important as kinases in regulatory processes.

Unit Definition

One unit will hydrolyze 1.0 μmole of p-nitrophenyl phosphate per min at pH 4.8 at 37 °C.

Physical form

Suspension in 1.8 M (NH4)2SO4, 10 mM MgCl2, pH 5.3

Pictograms

Health hazard

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Resp. Sens. 1

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Susumu Yamato et al.
Biological & pharmaceutical bulletin, 27(2), 210-215 (2004-02-06)
Sweet potato acid phosphatase was covalently coupled with glutaraldehyde to aminopropyl controlled-pore glass, and used as a pre-column enzyme reactor. The immobilized enzyme reactor (IMER) was continuously operated using an automated chromatographic detection system we developed. Functional evaluation of the
G Schenk et al.
Archives of biochemistry and biophysics, 370(2), 183-189 (1999-10-08)
Purple acid phosphatases comprise a family of binuclear metal-containing acid hydrolases, representatives of which have been found in animals, plants, and fungi. The goal of this study was to characterize purple acid phosphatases from sweet potato tubers and soybean seeds
H Kawabe et al.
Biochimica et biophysica acta, 784(1), 81-89 (1984-01-18)
The function of Mn(III) in plant acid phosphatase has been investigated by a metal-substitution study, and some properties of the Fe(III)-substituted enzyme were compared with those of the native Mn(III) enzyme and mammalian Fe(III)-containing acid phosphatases. 19F nuclear magnetic resonance
S M Duff et al.
Proceedings of the National Academy of Sciences of the United States of America, 88(21), 9538-9542 (1991-11-01)
Induction of phosphatase activity is an important component of the plant cell response to phosphate deficiency. Suspension cell cultures of Brassica nigra contain two major inducible acid phosphatase (APase) isozymes; vacuolar phosphoenolpyruvate (PEP) APase and cell wall nonspecific APase. Polyclonal
An unstable manganese(III) complex incorporating ligand donor types proposed for an acid phosphatase from sweet potato: (p-nitrobenzenethiolato)[N,N'-ethylenebis(salicylideneaminato)]manganese(III)
Gohdes, J. and W. Armstrong
Inorganic Chemistry, 27, 1841-1842 (1988)

Articles

Instructions for working with enzymes supplied as ammonium sulfate suspensions

Protocols

Enzymatic Assay of Acid Phosphatase (EC 3.1.3.2)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service