Skip to Content
Merck
All Photos(3)

Key Documents

P1146

Sigma-Aldrich

Phosphatase, Acid from potato

lyophilized powder, ≥3.0 units/mg solid

Synonym(s):

(Orthophosphoric-monoester phosphohydrolase acid optimum), Acid Phosphatase from potato

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
Enzyme Commission number:
EC Number:
MDL number:
UNSPSC Code:
12352204
eCl@ss:
42010102
NACRES:
NA.54

form

lyophilized powder

Quality Level

specific activity

≥3.0 units/mg solid

mol wt

69 kDa

shipped in

wet ice

storage temp.

−20°C

SMILES string

O(CC(O)C)CC#C

InChI

1S/C6H10O2/c1-3-4-8-5-6(2)7/h1,6-7H,4-5H2,2H3

InChI key

GZCWLCBFPRFLKL-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Acid phosphatase from potato is a phosphomonoesterase, which can appear in multiple molecular forms of similar molecular mass but with different isoelectric points.

Application

Acid phosphatase from potato has been used in a study to assess the potential allergenicity of novel gene products. It has also been used in a study to remove eight phosphate groups from casein at a pH of 7.0.
The activity of potato acid phosphatase in various surfactant medias was examined. Bis(2- ethylhexyl)sodium sulfosuccinate, Brij 35, and SDS at 3mM inhibited phosphatase activity while the surfactant cetyltrimethylammonium bromide enhanced activity.

Biochem/physiol Actions

Acid phosphatases (APase) are a family of enzymes that non-specifically catalyze the hydrolysis of monoesters and anhydrides of phosphoric acid to produce inorganic phosphate at an optimum pH of 4 to 7.

Unit Definition

One unit will hydrolyze 1.0 μmole of p-nitrophenyl phosphate per min at pH 4.8 at 37 °C.

Pictograms

Health hazard

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Resp. Sens. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

J Lalitha et al.
Biochemistry and molecular biology international, 41(4), 797-803 (1997-04-01)
The effect of three different classes of surfactants viz., anionic, cationic and neutral on catalytic activity of potato acid phosphatase (AcPase) was studied. Anionic surfactants bis(2-ethylhexyl) sodium sulfosuccinate (AOT) and sodium dodecyl sulfate (SDS) inhibited AcPase activity completely at 3
P P Waymack et al.
Archives of biochemistry and biophysics, 288(2), 621-633 (1991-08-01)
An acid phosphatase (orthophosphoric monoester phosphohydrolase, acid optimum; EC 3.1.3.2) isoenzyme from wheat germ was purified 7000-fold to homogeneity. The effect of wheat germ sources and their relationship to the isoenzyme content and purification behavior of acid phosphatases was investigated.
Mariusz Olczak et al.
Acta biochimica Polonica, 50(4), 1245-1256 (2004-01-24)
The properties of plant purple acid phosphatases (PAPs), metallophosphoesterases present in some bacteria, plants and animals are reviewed. All members of this group contain a characteristic set of seven amino-acid residues involved in metal ligation. Animal PAPs contain a binuclear
Y Sugiura et al.
The Journal of biological chemistry, 256(20), 10664-10670 (1981-10-25)
A new manganese-containing acid phosphatase has been isolated and crystallized from sweet potato tubers. The pure enzyme contains one atom of manganese per Mr = 110,000 polypeptide and shows phosphatase activity toward various phosphate substrates. The pH optimum of the
S I Tu et al.
Plant physiology, 88(1), 61-68 (1988-09-01)
Primary cell walls, free from cytoplasmic contamination were prepared from corn (Zea mays L.) roots and potato (Solanum tuberosum) tubers. After EDTA treatment, the bound acid phosphatase activities were measured in the presence of various multivalent cations. Under the conditions

Protocols

Enzymatic Assay of Acid Phosphatase (EC 3.1.3.2)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service