Skip to Content
Merck

Skip To

F7253

Fast Black K Salt hemi(zinc chloride) salt

Dye content, ≥25%, practical grade, powder

Synonym(s):

2,5-Dimethoxy-4-([4-nitrophenyl]azo)benzenediazonium chloride hemi(zinc chloride) salt, Brentamine fast black K salt

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Pack SizeSKUAvailabilityPrice

About This Item

Linear Formula:
C14H12N5O4 · 1/2ZnCl4
CAS Number:
Molecular Weight:
417.88
Colour Index Number:
37190
UNSPSC Code:
12171500
PubChem Substance ID:
EC Number:
264-656-1
NACRES:
NA.47
MDL number:
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Product Name

Fast Black K Salt hemi(zinc chloride) salt, practical grade

grade

practical grade

Quality Segment

form

powder

composition

Dye content, ≥25%

technique(s)

microbe id | staining: suitable

color

brown-red

mp

150 °C

solubility

water: 10 mg/mL

application(s)

diagnostic assay manufacturing
hematology
histology

storage temp.

room temp

SMILES string

[Cl-].[Cl-].Cl[Zn]Cl.COc1cc([N+]#N)c(OC)cc1N=Nc2ccc(cc2)[N+]([O-])=O.COc3cc([N+]#N)c(OC)cc3N=Nc4ccc(cc4)[N+]([O-])=O

InChI

1S/2C14H12N5O4.4ClH.Zn/c2*1-22-13-8-12(14(23-2)7-11(13)16-15)18-17-9-3-5-10(6-4-9)19(20)21;;;;;/h2*3-8H,1-2H3;4*1H;/q2*+1;;;;;+2/p-4

InChI key

NZWZSUSPTZUGNN-UHFFFAOYSA-J

General description

Fast black K (C.l. 37190) is a lipophilic amino-substituted monoazo dye. It is also known as Brentamine fast black K salt.

Application

Fast Black K Salt hemi (zinc chloride) salt has been used for the staining of acid phosphatase activity in SDS-PAGE (sodium dodecyl sulfate polyacrylamide gel electrophoresis) and native-PAGE. It has been used for staining endopeptidase and leucine aminopeptidase activity.

Fast black K salt has various applications as listed below-

  • to stain enterochromaffin cells > as an indirect stain for periodate-generated tissue aldehyde groups
  • as a visualization agent for amphetamines and cyanobacterial toxins following their separation by thin-layer chromatography
  • for the staining of acid phosphatase activity in SDS-PAGE (sodium dodecyl sulfate polyacrylamide gel electrophoresis) and native-PAGE
  • staining endopeptidase and leucine aminopeptidase activity

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
F337844670F3381
grade

practical grade

grade

-

grade

-

grade

-

technique(s)

microbe id | staining: suitable

technique(s)

microbe id | staining: suitable

technique(s)

microbe id | staining: suitable

technique(s)

-

color

brown-red

color

-

color

-

color

-

solubility

water: 10 mg/mL

solubility

H2O: 10 mg/mL

solubility

hot water: 0.01 g/10 mL

solubility

H2O: 1 mg/mL

form

powder

form

powder

form

powder

form

powder

application(s)

diagnostic assay manufacturing
hematology
histology

application(s)

diagnostic assay manufacturing
hematology
histology

application(s)

diagnostic assay manufacturing
hematology
histology

application(s)

diagnostic assay manufacturing
hematology
histology


pictograms

Exclamation mark

signalword

Warning

hcodes

Hazard Classifications

Eye Irrit. 2

Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

dust mask type N95 (US), Eyeshields, Gloves



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library


Related Content


Genetic control of isozymes in the primary gene pool Phaseolus lunatus L.
Zoro BI
Biotechnologie, agronomie, societe et environnement, 3, 10-27 (1999)
Analyses of Root-secreted Acid Phosphatase Activity in Arabidopsis.
Wang L and Liu D
Bio-protocol null



Global Trade Item Number

SKUGTIN
F7253-25G04061835347711
F7253-50G04061833619414
F7253-100G04061835347704

Questions

Reviews

No rating value

Active Filters