Skip to Content
Merck
All Photos(1)

Documents

44670

Sigma-Aldrich

Fast Blue BB Salt hemi(zinc chloride) salt

for microscopy (Hist.)

Synonym(s):

4-Amino-2,5-diethoxybenzanilide diazotated zinc double salt, 4-Benzoylamino-2,5-diethoxybenzenediazonium chloride hemi(zinc chloride) salt

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C17H18N3O3Cl · 1/2 ZnCl2
CAS Number:
Molecular Weight:
415.94
Colour Index Number:
37175
Beilstein:
1845288
EC Number:
MDL number:
UNSPSC Code:
12171500
PubChem Substance ID:
NACRES:
NA.47

grade

for microscopy (Hist.)

form

powder

mp

157 °C

solubility

hot water: 0.01 g/10 mL

εmax

≥100 at 390-400 nm in water
≥200 at 325-335 nm in water

application(s)

diagnostic assay manufacturing
hematology
histology

storage temp.

room temp

SMILES string

[Cl-].[Cl-].Cl[Zn]Cl.CCOc1cc([N+]#N)c(OCC)cc1NC(=O)c2ccccc2.CCOc3cc([N+]#N)c(OCC)cc3NC(=O)c4ccccc4

InChI

1S/2C17H17N3O3.4ClH.Zn/c2*1-3-22-15-11-14(20-18)16(23-4-2)10-13(15)19-17(21)12-8-6-5-7-9-12;;;;;/h2*5-11H,3-4H2,1-2H3;4*1H;/q;;;;;;+2/p-2

InChI key

CMFRFQODFZBKTI-UHFFFAOYSA-L

Looking for similar products? Visit Product Comparison Guide

General description

Fast blue BB salt hemi(zinc chloride) salt is suitable as a common coloring agent for TLC (thin layer chromatography) analysis of cannabis. It is also suitable for histochemistry studies. It is of 415.9kDa.

Application

Fast blue BB salt hemi(zinc chloride) salt has been used to prepare substrate solution for tissue-blot enzyme-immunoassay procedure. It has been used in immunohistochemical staining.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

L Wagner et al.
Journal of immunological methods, 142(2), 147-155 (1991-09-13)
A trypsin-like serine esterase (SE) is known to be present in cultured cells with cytolytic potential. The distribution pattern of this enzyme in haematological cells and body tissues has been assessed using a method which permits rapid identification of individual
Marjorie B Medina
Journal of agricultural and food chemistry, 59(5), 1565-1571 (2011-02-12)
A new method for the detection of phenolics in food systems was developed. This method is based on interactions of phenolics with Fast Blue BB diazonium salt in alkali pH, forming azo complexes, with the absorbance measured at 420 nm
B Algermissen et al.
Experimental dermatology, 3(6), 290-297 (1994-12-01)
In order to gain insights into the dynamics of mast cell subpopulations in normal and diseased skin, a novel enzyme-histochemical double and triple staining method was employed that allowed the detection of metachromasia (toluidine blue) and the mast cell proteases
I Ojanperä et al.
Journal of analytical toxicology, 12(2), 108-110 (1988-03-01)
A sensitive visual method for the thin-layer chromatography of beta-adrenergic blocking drugs is described. The Rf values and the detection limits of eleven beta-blockers extracted from urine and as pure drugs are given. Other commonly used cardiovascular drugs are shown
L H Rademakers et al.
The Histochemical journal, 21(5), 301-308 (1989-05-01)
Non-specific esterase (NSE) activity was demonstrated in glutaraldehyde-fixed monolayers of murine peritoneal macrophages. Using 2-naphthylthiol acetate (NTA) as substrate and Fast Blue BB as coupling agent a strong osmiophilic reaction product was obtained. The reaction product was observed as electron-dense

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service