Skip to Content
Merck

Skip To

F6428

Free Glycerol Reagent

used for quantitative enzymatic determination of glycerol

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Pack SizeSKUAvailabilityPrice

About This Item

MDL number:
UNSPSC Code:
12161503
NACRES:
NA.25
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Level

form

solid

usage

40 mL sufficient for 50 reactions

color

white to off-white

functional group

hydroxyl

relevant disease(s)

obesity

storage temp.

2-8°C

SMILES string

C(C(CO)O)O

InChI

1S/C3H8O3/c4-1-3(6)2-5/h3-6H,1-2H2

InChI key

PEDCQBHIVMGVHV-UHFFFAOYSA-N

General description

The Free Glycerol Reagent provides a precise method for measuring free endogenous glycerol levels through coupled enzyme reactions, bypassing the need for initial lipase hydrolysis. Unlike triglycerides, which are typically bound to proteins and transported in plasma, free glycerol circulates independently. The innovative approach involves enzymatic or alkaline hydrolysis of triglycerides, liberating glycerol and free fatty acids for accurate measurement.

The Serum Triglyceride Determination Kit (Catalog Number TR0100) offers a comprehensive solution for assessing glycerol, true triglycerides, or total triglycerides in serum or plasma samples. Utilizing lipase enzymatic hydrolysis, triglycerides are converted into glycerol and free fatty acids, enabling precise quantification through coupled enzyme reactions by F6428.

Unlike many commercially available triglyceride reagents, this method distinguishes between endogenous glycerol and glycerol derived from lipase hydrolysis of glycerides. With this advanced technology, researchers can confidently analyze glycerol levels with exceptional accuracy and reliability.

Application

The Free Glycerol Reagent facilitates the quantitative enzymatic determination of glycerol levels in serum or plasma. Additionally, it may also be used to measure glycerol release from lipolysis in adipose tissue.

Biochem/physiol Actions

Glycerol undergoes phosphorylation through the enzymatic action of glycerol kinase (GK), utilizing adenosine-5′-triphosphate (ATP) to produce glycerol-1-phosphate (G-1-P) and adenosine-5′-diphosphate (ADP). Subsequently, glycerol-1-phosphate (G-1-P) is oxidized by glycerol phosphate oxidase (GPO) to yield dihydroxyacetone phosphate (DAP) and hydrogen peroxide (H2O2). This process involves peroxidase (POD), which catalyzes the coupling of hydrogen peroxide (H2O2) with 4-aminoantipyrine (4-AAP) and sodium N-ethyl-N-(3-sulfopropyl) m-anisidine (ESPA) to generate a quinoneimine dye exhibiting an absorbance maximum at 540 nm.The observed increase in absorbance at 540 nm correlates directly with the concentration of free glycerol present in the sample. This method provides a reliable means for quantifying free glycerol levels, enabling accurate assessment of biochemical samples.
The free glycerol reagent uses coupled enzyme reactions resulting in an increase in absorbance at 540 nm that is directly proportional to the glycerol concentration.

Features and Benefits

  • Simplified Process: Streamline your workflow with a single reagent, offering a quick 5-minute reaction time.
  • User-Friendly Design: Eliminate the need for weighing or mixing multiple reagents, ensuring ease of use and efficiency.

Other Notes

For additional information on our range of Biochemicals, please complete this form.
In addition to kits, the individual reagents and glycerol standard are available separately when fewer reactions are needed.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
G7793FG0100374813
functional group

hydroxyl

functional group

-

functional group

-

functional group

-

storage temp.

2-8°C

storage temp.

2-8°C

storage temp.

2-8°C

storage temp.

2-8°C

form

solid

form

-

form

-

form

liquid

color

white to off-white

color

-

color

-

color

-

Quality Level

200

Quality Level

200

Quality Level

200

Quality Level

100

relevant disease(s)

obesity

relevant disease(s)

-

relevant disease(s)

-

relevant disease(s)

-


signalword

Danger

Storage Class

11 - Combustible Solids

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

dust mask type N95 (US), Eyeshields, Gloves

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - ED ENV 1 - Eye Dam. 1 - Skin Irrit. 2



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library


Related Content


Geonho Kim et al.
Journal of insect physiology, 106(Pt 1), 13-19 (2017-05-20)
Acetic acid is a fermentation product of many microorganisms, including some that inhabit the food and guts of Drosophila. Here, we investigated the effect of dietary acetic acid on oviposition and larval performance of Drosophila. At all concentrations tested (0.34-3.4%)
Adipocyte Gs but not Gi signaling regulates whole-body glucose homeostasis
Caron A, et al.
Molecular Metabolism, 27, 11-21 (2019)
Claire Regazzetti et al.
Diabetes, 58(1), 95-103 (2008-11-06)
Obesity is characterized by an overgrowth of adipose tissue that leads to the formation of hypoxic areas within this tissue. We investigated whether this phenomenon could be responsible for insulin resistance by studying the effect of hypoxia on the insulin



Global Trade Item Number

SKUGTIN
F6428-40ML04061835544899

Questions

1–5 of 5 Questions  
  1. How can I determine the shelf life / expiration / retest date of this product?

    1 answer
    1. If this product has an expiration or retest date, it will be shown on the Certificate of Analysis (COA, CofA). If there is no retest or expiration date listed on the product's COA, we do not have suitable stability data to determine a shelf life. For these products, the only date on the COA will be the release date; a retest, expiration, or use-by-date will not be displayed.
      For all products, we recommend handling per defined conditions as printed in our product literature and website product descriptions. We recommend that products should be routinely inspected by customers to ensure they perform as expected.
      For products without retest or expiration dates, our standard warranty of 1 year from the date of shipment is applicable.
      For more information, please refer to the Product Dating Information document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/418/501/product-dating-information-06-25-mk.pdf

      Helpful?

  2. I would like to measure the concentration of free glycerol in the culture supernatant of adipocytes using this reagent with a plate reader instead of a cuvette. What are recommended sample and reagent volumes for a plate reader? Have you ever tested?

    1 answer
    1. This product has not been tested for use with a well plate format. The product instructions call for a total volume of 830 µL, where 10 µL each of water, glycerol standard, and sample are added to cuvettes containing 800 µL of Free Glycerol Reagent. Most 96-well plates have a working volume of 100-300 µL, so the volume of each component would need to be scaled accordingly. Adding 3 µL each of water, glycerol standard, and sample to wells containing 240 µL of Free Glycerol Reagent would result in the same concentrations with a total volume of 249 µL. Regardless, the functionality of a plate reader in this application cannot be guaranteed and would need to be assessed independently.

      Helpful?

  3. I have an old version of this product (6428) - just one bottle not a kit. I need to reconstitute this but can't find instructions. Can you let me know what to reconstitute in and how much? Thank you!

    1 answer
    1. There are no specific instructions for 6428. F6428 is one bottle of reagent as well. The user guide of F6428 may serve as a reference. A typical reconstitution procedure of F6428 is: Reconstitute the Free Glycerol Reagent with 40 ml of water. After addition of water, stopper the vials, and immediately mix several times by inversion. DO NOT SHAKE. Protect the reagent from light by storing in amber bottles. Please check page 2 of the user guide here:
      https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/product/documents/352/968/f6428bul.pdf

      Helpful?

  4. How is shipping temperature determined? And how is it related to the product storage temperature?

    1 answer
    1. Products may be shipped at a different temperature than the recommended long-term storage temperature. If the product quality is sensitive to short-term exposure to conditions other than the recommended long-term storage, it will be shipped on wet or dry-ice. If the product quality is NOT affected by short-term exposure to conditions other than the recommended long-term storage, it will be shipped at ambient temperature. As shipping routes are configured for minimum transit times, shipping at ambient temperature helps control shipping costs for our customers. For more information, please refer to the Storage and Transport Conditions document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/316/622/storage-transport-conditions-mk.pdf

      Helpful?

  5. Could you kindly send me a package insert and a manual application for determination on automated clinical systems? Many thanks.

    1 answer
    1. The Free Glycerol Reagent is for R&D use only, not for in vitro diagnostic use, drug, household, or other uses. Please consult the Material Safety Data Sheet for information regarding hazards and safe handling practices.

      Helpful?

Reviews

No rating value

Active Filters