G7793
Glycerol Standard Solution
2.5 mg/ml equivalent triolein concentration
Synonym(s):
Triolein Standard Solution
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
UNSPSC Code:
41116107
NACRES:
NA.25
Recommended Products
Application
The triglyceride and free glycerol kits are for the quantitative determination of glycerol, total triglycerides or free triglycerides.
Linkage
In addition to kits, the individual reagents and glycerol standard are available separately when fewer reactions are needed.
required but not provided
Product No.
Description
Pricing
Storage Class Code
12 - Non Combustible Liquids
WGK
WGK 1
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Sarah-Jane Leigh et al.
Nutrition & metabolism, 17, 71-71 (2020-08-25)
Exercise has been extensively utilised as an effective therapy for overweight- and obesity-associated changes that are linked to health complications. Several preclinical rodent studies have shown that treadmill exercise alongside an unhealthy diet improves metabolic health and microbiome composition. Furthermore
Neda Barghi et al.
PLoS biology, 17(2), e3000128-e3000128 (2019-02-05)
The genetic architecture of adaptive traits is of key importance to predict evolutionary responses. Most adaptive traits are polygenic-i.e., result from selection on a large number of genetic loci-but most molecularly characterized traits have a simple genetic basis. This discrepancy
Mingyu Shin et al.
Frontiers in immunology, 11, 63-63 (2020-02-23)
Drosophila hemocytes, like those of mammals, are given rise from two distinctive phases during both the embryonic and larval hematopoiesis. Embryonically derived hemocytes, mostly composed of macrophage-like plasmatocytes, are largely identified by genetic markers. However, the cellular diversity and distinct
H Y Tan et al.
The Journal of physiology, 594(24), 7309-7326 (2016-08-26)
Loss of function of the melanocortin 4 receptor (MC4R) results in hyperphagia, obesity and increased growth. Despite knowing that MC4Rs control food intake, we are yet to understand why defects in the function of the MC4R receptor contribute to rapid
Christian Baumeier et al.
Molecular metabolism, 6(10), 1254-1263 (2017-10-17)
Increased hepatic expression of dipeptidyl peptidase 4 (DPP4) is associated with non-alcoholic fatty liver disease (NAFLD). Whether this is causative for the development of NAFLD is not yet clarified. Here we investigate the effect of hepatic DPP4 overexpression on the
Articles
Lipid Induced Insulin Resistance
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service