Skip to Content
Merck

Skip To

F6136

Anti-Horse Spleen Ferritin antibody produced in rabbit

affinity isolated antibody, lyophilized powder

Synonym(s):

Ferritin Antibody

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Pack SizeSKUAvailabilityPrice

About This Item

UNSPSC Code:
12352203
NACRES:
NA.46
MDL number:
Conjugate:
unconjugated
Clone:
polyclonal
Application:
ELISA (i), IEP
Citations:
31
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


biological source

rabbit

Quality Segment

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

lyophilized powder

species reactivity

horse

technique(s)

immunoelectrophoresis: suitable, indirect ELISA: 1:10,000

storage temp.

2-8°C

target post-translational modification

unmodified

Gene Information

horse ... Ftl(100051593)

General description

Ferritin consists of homogenous protein segment called as apoprotein and an iron segment which binds to the apoprotein. It is composed of heavy (H) and light (L) chains.
Ferritin is the storage form of iron in mammals, found mainly in liver, bone marrow and spleen. It is also present in small intestine, placenta, kidney and heart. Horse spleen ferritin is made up of 24 identical subunits. Ferritin of different organs is distinct with different electrophoretic mobility and possible functional differences.
.

Immunogen

Purified horse spleen ferritin

Application

Anti-Horse Spleen Ferritin antibody was used for immunocytochemistry of rat brain sections at a working antibody dilution of 1:2000 and in mouse brain sections at 1:1000. The antibody was used for immunoblotting using cell lysates of mouse primary erythroblasts and human ovarian carcinoma cells and human bronchial epithelial cells at a working dilution of 1:1000. The antibody is suitable for immunoelectrophoresis.
Horse Spleen Ferritin antibody has been used:
  • immunofluorescence
  • in western blotting
  • in immunohistochemistry

Biochem/physiol Actions

Ferritin iron is oxidized from Fe(II) to Fe(III) in the presence of the enzyme ferroxidase.
The antibody specifically recognizes ferritin from horse spleen.

Physical form

Lyophilized from 0.01 M sodium phosphate, 0.015 M sodium chloride, pH 7.2

Preparation Note

Reconstitute with 0.135 M sodium chloride.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
F5012H8890A9521
Quality Level

200

Quality Level

200

Quality Level

200

Quality Level

200

antibody form

affinity isolated antibody

antibody form

whole antiserum

antibody form

whole antiserum

antibody form

IgG fraction of antiserum

conjugate

unconjugated

conjugate

unconjugated

conjugate

unconjugated

conjugate

unconjugated

biological source

rabbit

biological source

rabbit

biological source

rabbit

biological source

rabbit

clone

polyclonal

clone

polyclonal

clone

polyclonal

clone

polyclonal

Gene Information

horse ... Ftl(100051593)

Gene Information

human ... FTH1(2495)

Gene Information

-

Gene Information

-


Still not finding the right product?

Try our Product Selector Tool to narrow your options


Storage Class

11 - Combustible Solids

wgk

WGK 2

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library


Related Content






Global Trade Item Number

SKUGTIN
F6136-1MG04061833618486

Questions

Reviews

No rating value

Active Filters