Skip to Content
Merck
All Photos(2)

Key Documents

F5012

Sigma-Aldrich

Anti-Ferritin, Human antibody produced in rabbit

whole antiserum, liquid

Synonym(s):

Anti-FHC, Anti-FTH, Anti-FTHL6, Anti-HFE5, Anti-PIG15, Anti-PLIF

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203
NACRES:
NA.46
Pricing and availability is not currently available.

biological source

rabbit

conjugate

unconjugated

antibody form

whole antiserum

antibody product type

primary antibodies

clone

polyclonal

form

liquid

contains

15 mM sodium azide

species reactivity

human

technique(s)

Ouchterlony double diffusion: 1:16

UniProt accession no.

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... FTH1(2495)

General description

Ferritin, an iron-storage protein is usually present in the liver and spleen in mammals. Iron and ferritin are distributed relatively same in the eye. Ferritin is made up of heavy and light chains.

Immunogen

Ferritin isolated from normal human spleen

Application

  • Anti-Ferritin, Human antibody produced in rabbit has been used in:
  • immunohistochemistry
  • western blotting
  • immunocytochemical techniques

Anti-Ferritin, Human antibody is suitable for use in Ouchterlony double diffusion (1:16 using human spleen ferritin), and immunoelectrophoresis. The antibody may also be used in western blot (1:1000 using frozen liver homogenates) and immunoprecipitation (using 300 μg of cell lysate incubated with 6 mul of anti-ferritin for 1h at 4 ° C).
Applications in which this antibody has been used successfully, and the associated peer-reviewed papers, are given below.
Western Blotting (1 paper)

Biochem/physiol Actions

Ferritin is an iron-storage protein that regulates iron-homeostasis in cells. While mitochondrial ferritin is a homopolymer, the cytosolic form consists of heteropolymeric L- and H-chains. Genetic mutations in ferritin L-chains have been associated with neurodegenerative disorders, whereas altered levels of ferritin have been linked to hemophagocytic lymphohistiocytosis and adult-onset Still′s disease . In immunoelectrophoresis, anti-ferritin, human antibody is specific for ferritin when used against human spleen extract and purified ferritin. No reactivity is observed against normal human serum.

Preparation Note

The antiserum has been treated to remove lipoproteins.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Scientific tests
Aldred EM, et al.
Pharmacology, 331-341 (2009)
Increased Iron Sequestration in Alveolar Macrophages in Chronic Obtructive Pulmonary Disease
Philippot Q, et al.
Testing, 9(5), e96285-e96285 (2014)
Moon-Suhn Ryu et al.
The Journal of clinical investigation, 127(5), 1786-1797 (2017-04-05)
Developing erythrocytes take up exceptionally large amounts of iron, which must be transferred to mitochondria for incorporation into heme. This massive iron flux must be precisely controlled to permit the coordinated synthesis of heme and hemoglobin while avoiding the toxic
Pro-inflammatory activation of microglia in the brain of patients with sepsis
Zrzavy T, et al.
Neuropathology and Applied Neurobiology, 45(3), 278-290 (2019)
Zhao Wang et al.
International journal of hematology, 90(4), 501-505 (2009-08-06)
Elevated levels of serum ferritin and a low percentage of glycosylated ferritin have been reported in adult-onset Still's disease (AOSD) as well as in hemophagocytic lymphohistiocytosis (HLH). The objective of the current study was to investigate total ferritin levels and

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service