Skip to Content
Merck
All Photos(3)

Key Documents

E1752

Sigma-Aldrich

Estrone 3-(β-D-glucuronide) sodium salt

Synonym(s):

1,3,5(10)-Estratrien-17-one 3-glucuronide sodium salt

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C24H29NaO8
CAS Number:
Molecular Weight:
468.47
Beilstein:
6043458
MDL number:
UNSPSC Code:
51111800
PubChem Substance ID:
NACRES:
NA.77

sterility

non-sterile

Assay

≥99% (HPLC)

form

powder

technique(s)

ELISA: suitable

solubility

water: 20 mg/mL, clear, colorless

shipped in

ambient

storage temp.

−20°C

SMILES string

[Na+].[H][C@]12CC[C@]3(C)C(=O)CC[C@@]3([H])[C@]1([H])CCc4cc(O[C@@H]5O[C@@H]([C@@H](O)[C@H](O)[C@H]5O)C([O-])=O)ccc24

InChI

1S/C24H30O8.Na/c1-24-9-8-14-13-5-3-12(10-11(13)2-4-15(14)16(24)6-7-17(24)25)31-23-20(28)18(26)19(27)21(32-23)22(29)30;/h3,5,10,14-16,18-21,23,26-28H,2,4,6-9H2,1H3,(H,29,30);/q;+1/p-1/t14-,15-,16+,18+,19+,20-,21+,23-,24+;/m1./s1

InChI key

PBULYLQKWDXDFZ-QHIGSJCSSA-M

Looking for similar products? Visit Product Comparison Guide

Application

Estrone 3-(β-D-glucuronide) sodium salt has been used as a standard:
  • to estimate estrone conjugate (E1Cs) from urine samples by enzyme immunoassays (EIAs)
  • to determine corticosterone metabolites from fecal and water samples by EIA
  • to estimate progesterone and estrogen levels in feces of Chinese goral by EIA

Biochem/physiol Actions

Estrone 3 glucuronide (E3G) is a urinary metabolite of estrogen. Higher levels of estrone 3 glucuronide are observed during the early onset of the fertile period of the menstrual cycle.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Rebecca C Miller et al.
Clinical chemistry, 50(5), 924-932 (2004-04-24)
Specific gravity (SG) may perform as well as creatinine (CR) correction for adjusting urinary hormone concentrations, as well as offer some advantages. We compared the two methods and applied them to US and Bangladeshi specimens to evaluate their use in
Lateral Flow and Consumer Diagnostics
Tiplady S
The Immunoassay Handbook, 533-536 null
Andrew H Nagel et al.
Conservation physiology, 7(1), coz074-coz074 (2019-11-19)
This study sought to develop non-invasive techniques to monitor glucocorticoids in captive Necturus as a means to correlate inter-renal gland function in relation to environmental and physiological changes. Six individually housed breeding pairs of captive Necturus beyeri were subjected to
Leonard F Blackwell et al.
Frontiers in public health, 6, 153-153 (2018-06-16)
The patterns of a woman's normal ovarian activity can take many forms from childhood to menopause. These patterns lie on a continuum ranging from no ovarian activity to a fully fertile ovulatory cycle, but among the other defined patterns are
Kathleen A O'Connor et al.
American journal of physical anthropology, 145(4), 629-638 (2011-06-28)
Psychosocial stress is thought to negatively impact fecundity, but human studies are confounded by variation in nutrition and lifestyle. Baboons offer a useful model to test the effect of prolonged mild stress on reproductive indicators in a controlled setting. Following

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service