Skip to Content
Merck

E2127

β-Estradiol 3-(β-D-glucuronide) sodium salt

≥98% (HPLC), estradiol metbolite, powder

Synonym(s):

1,3,5(10)-Estratriene-3,17β-diol 3-glucuronide, 3,17β-Dihydroxy-1,3,5(10)-estratriene 3-glucuronide

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Linear Formula:
C24H31O8Na
CAS Number:
Molecular Weight:
470.49
UNSPSC Code:
51111800
PubChem Substance ID:
NACRES:
NA.77
MDL number:
Assay:
≥98% (HPLC)
Form:
powder
Quality level:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Product Name

β-Estradiol 3-(β-D-glucuronide) sodium salt,

sterility

non-sterile

Quality Level

assay

≥98% (HPLC)

form

powder

solubility

water: 10 mg/mL, clear to slightly hazy

shipped in

ambient

storage temp.

−20°C

SMILES string

[Na].CC12CCC3C(CCc4cc(OC5OC(C(O)C(O)C5O)C(O)=O)ccc34)C1CCC2O

InChI

1S/C24H32O8.Na.H/c1-24-9-8-14-13-5-3-12(10-11(13)2-4-15(14)16(24)6-7-17(24)25)31-23-20(28)18(26)19(27)21(32-23)22(29)30;;/h3,5,10,14-21,23,25-28H,2,4,6-9H2,1H3,(H,29,30);;

InChI key

JKYLXXZHJUQLMK-UHFFFAOYSA-N

Application

β-Estradiol 3-(β-D-glucuronide) sodium salt has been used in estrogen administration. It has also been used to determine the amount of two types of glucuronidated estradiols such as estradiol-3- α glucuronide and estradiol-17- β glucuronide.


pictograms

Exclamation mark

signalword

Warning

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral

Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

dust mask type N95 (US), Eyeshields, Gloves



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Yuichiro Sato et al.
Xenobiotica; the fate of foreign compounds in biological systems, 42(9), 823-829 (2012-03-23)
The aims of this study were to quantify absolute protein levels of uridine 5'-diphosphate-glucuronosyltransferases (UGTs) 1A1 and 2B7 in human liver microsomes (HLMs) and to investigate their correlation with marker activities. A quantification method for UGT1A1 and UGT2B7 in HLMs
Direct, complex effects of estrogens on basal forebrain cholinergic neurons
Bora S H, et al.
Experimental Neurology, 194(2), 506-522 (2005)
Linda B Tabas et al.
Analytical biochemistry, 310(1), 61-66 (2002-11-05)
This study investigated a high-throughput assay to measure multidrug resistance-associated protein (MRP1)-mediated uptake into membrane vesicles. Typically, a rapid filtration technique using a 12-filter vacuum manifold is used. We report here the development of a 96-well microtiter dish assay. MRP1-transfected



Global Trade Item Number

SKUGTIN
E2127-25MG04061832849812
E2127-5MG04061833602454
E2127-10MG04061833602447