Skip to Content
Merck

Skip To

C9012

Bis(cyclohexanone)oxaldihydrazone

≥95% purity (TLC), powder

Synonym(s):

Cuprizon, Cuprizone, Oxalic acid bis(cyclohexylidenehydrazide)

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Pack SizeSKUAvailabilityPrice

About This Item

Empirical Formula (Hill Notation):
C14H22N4O2
CAS Number:
Molecular Weight:
278.35
NACRES:
NA.47
PubChem Substance ID:
UNSPSC Code:
12171500
EC Number:
206-729-2
MDL number:
Beilstein/REAXYS Number:
2388004
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Product Name

Bis(cyclohexanone)oxaldihydrazone, ≥95% (TLC)

Quality Level

assay

≥95% (TLC)

form

powder

technique(s)

titration: suitable

color

white/clear to off-white

mp

210-214 °C (lit.)

solubility

acetic acid: 50 mg/mL, colorless to very faintly brown

application(s)

diagnostic assay manufacturing
hematology
histology

storage temp.

2-8°C

SMILES string

O=C(N\N=C1\CCCCC1)C(=O)N\N=C2/CCCCC2

InChI

1S/C14H22N4O2/c19-13(17-15-11-7-3-1-4-8-11)14(20)18-16-12-9-5-2-6-10-12/h1-10H2,(H,17,19)(H,18,20)

InChI key

DSRJIHMZAQEUJV-UHFFFAOYSA-N

Application

  • Bis(cyclohexanone)oxaldihydrazone (Cuprizone) has been used in mice to study the effect of NG2 (neural/glial antigen 2) proteoglycan on toxic-induced demyelination.[1]
  • It has been used to induce demyelination in mice.
  • It has been used to study cuprizone-induced demyelination and remyelination in the corpus callosum of mice.

Biochem/physiol Actions

Bis(cyclohexanone)oxaldihydrazone (Cuprizone) is a copper chelator. It results in oligodendroglial cell death, thereby leading to demyelination, astrogliosis and microglia activation.[1] Cuprizone is widely used to study mitochondrial dysfunction-associated primary demyelination and remyelination in the corpus callosum.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
14690E3257D5143
technique(s)

titration: suitable

technique(s)

UV/Vis spectroscopy: suitable

technique(s)

-

technique(s)

-

color

white/clear to off-white

color

-

color

white to off-white

color

white to yellow

solubility

acetic acid: 50 mg/mL, colorless to very faintly brown

solubility

-

solubility

acetic acid: water (1:1): ≤50 mg/mL, DMF: soluble

solubility

1 M HCl: 50 mg/mL, clear to very slightly hazy

application(s)

diagnostic assay manufacturing
hematology
histology

application(s)

-

application(s)

diagnostic assay manufacturing
hematology
histology

application(s)

diagnostic assay manufacturing
hematology
histology

form

powder

form

powder

form

powder

form

powder or crystals

storage temp.

2-8°C

storage temp.

-

storage temp.

2-8°C

storage temp.

2-8°C


Still not finding the right product?

Explore all of our products under Bis(cyclohexanone)oxaldihydrazone


Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Cortical demyelination is prominent in the murine cuprizone model and is strain-dependent.
Skripuletz T, et al.
The American Journal of Pathology, 172, 1053-1061 (2008)
Demyelination and remyelination in anatomically distinct regions of the corpus callosum following cuprizone intoxication.
Steelman AJ, et al.
Neuroscience Research, 72, 32-42 (2012)
Galanin transgenic mice with elevated circulating galanin levels alleviate demyelination in a cuprizone-induced MS mouse model.
Zhang L, et al.
PLoS ONE, 7, e33901-e33901 (2012)



Global Trade Item Number

SKUGTIN
C9012-25G04061835347339

Questions

  1. Is it necessary to work in a fume hood when handling this chemical?

    1 answer
    1. This product does not require handling under a fume hood.

      Helpful?

Reviews

No rating value

Active Filters