Skip to Content
AE
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Bis(cyclohexanone)oxaldihydrazone
Synonym(s):
Cuprizon, Cuprizone, Oxalic acid bis(cyclohexylidenehydrazide)
Empirical Formula (Hill Notation):
C
14
H
22
N
4
O
2
CAS Number:
370-81-0
Molecular Weight:
278.35
EC Number:
206-729-2
Products
Product Category
stains (1)
Brand
Sigma-Aldrich (1)
Supelco (1)
Color
Formula Weight
Melting Point (°C)
Physical Form
Purity
Quality Segment
Application
Technique
Filter Products
Showing 1-2 of 2
Compare
Product Number
Description
Pricing
C9012
Bis(cyclohexanone)oxaldihydrazone,
≥95% purity (TLC), powder
Expand
View Pricing
14690
Bis(cyclohexanone)oxaldihydrazone,
for spectrophotometric det. of Cu, ≥99.0%
Expand
View Pricing