Skip to Content
Merck

Skip To

03416

Thymoquinone

analytical standard

Synonym(s):

2-Isopropyl-5-methyl-1,4-benzoquinone

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Pack SizeSKUAvailabilityPrice

About This Item

Empirical Formula (Hill Notation):
C10H12O2
CAS Number:
Molecular Weight:
164.20
NACRES:
NA.24
PubChem Substance ID:
UNSPSC Code:
85151701
EC Number:
207-721-1
MDL number:
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


grade

analytical standard

Quality Segment

assay

≥98.5% (GC)

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable, gas chromatography (GC): suitable

bp

230-232 °C (lit.)

mp

45-47 °C (lit.)

application(s)

cleaning products
cosmetics
flavors and fragrances
food and beverages
personal care

format

neat

SMILES string

CC(C)C1=CC(=O)C(C)=CC1=O

InChI

1S/C10H12O2/c1-6(2)8-5-9(11)7(3)4-10(8)12/h4-6H,1-3H3

InChI key

KEQHJBNSCLWCAE-UHFFFAOYSA-N

General description

Thymoquinone is employed in preclinical studies as neuroprotective and anticonvulsant agent.

Application

Thymoquinone has been used as an analytical standard for the determination of thymoquinone in Nigella sativa extracts and formulations by high performance thin layer chromatography (HPTLC). It may be used as a reference standard for the identification of thymoquinone as volatile aglycone in seasonally collected Satureja montana L. and in Juniperus oxycedrus L. by gas chromatography-mass spectrometry (GC-MS).

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
W306606W368008274666
technique(s)

HPLC: suitable, gas chromatography (GC): suitable

technique(s)

-

technique(s)

-

technique(s)

titration: suitable

application(s)

cleaning products
cosmetics
flavors and fragrances
food and beverages
personal care

application(s)

flavors and fragrances

application(s)

flavors and fragrances

application(s)

diagnostic assay manufacturing
hematology
histology

grade

analytical standard

grade

FG, Halal, Kosher

grade

FG, Fragrance grade, Halal, Kosher

grade

-

assay

≥98.5% (GC)

assay

-

assay

≥98%

assay

≥98%

format

neat

format

-

format

-

format

-

bp

230-232 °C (lit.)

bp

232 °C (lit.)

bp

161-162 °C (lit.)

bp

230-232 °C (lit.)


Still not finding the right product?

Explore all of our products under Thymoquinone


pictograms

Exclamation mark

signalword

Warning

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

target_organs

Respiratory system

Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

219.2 °F - closed cup

flash_point_c

104 °C - closed cup



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Gas chromatography mass spectral analysis of free and glycosidically bound volatile compounds from Juniperus oxycedrus L. growing wild in Croatia.
Milos, et al.
Food Chemistry, 68(3), 333-338 (2000)
Validated high-performance thin-layer chromatographic method for the quantification of thymoquinone in Nigella Sativa extracts and formulations.
Velho-Pereira, et al.
Phytochemical Analysis, 22(4), 367-373 (2011)
Stability-indicating ultra-performance liquid chromatography method for the estimation of thymoquinone and its application in biopharmaceutical studies.
Pathan, et al.
Biomedical Chromatography, 25(5), 613-620 (2011)



Global Trade Item Number

SKUGTIN
299669-10G04061826657614
299669-50G04061833596357
03416-100MG04061838618528

Questions

Reviews

No rating value

Active Filters