Skip to Content
Merck
All Photos(3)

Documents

274666

Sigma-Aldrich

Thymoquinone

≥98%

Synonym(s):

2-Isopropyl-5-methyl-1,4-benzoquinone

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H12O2
CAS Number:
Molecular Weight:
164.20
EC Number:
MDL number:
UNSPSC Code:
12171500
PubChem Substance ID:
NACRES:
NA.47

Assay

≥98%

form

solid

color

yellow

bp

230-232 °C (lit.)

mp

45-47 °C (lit.)

application(s)

diagnostic assay manufacturing
hematology
histology

storage temp.

room temp

SMILES string

CC(C)C1=CC(=O)C(C)=CC1=O

InChI

1S/C10H12O2/c1-6(2)8-5-9(11)7(3)4-10(8)12/h4-6H,1-3H3

InChI key

KEQHJBNSCLWCAE-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Thymoquinone (TQ), also known as 2-isopropyl-5-methyl-1, 4-benzoquinone, is present in a crystalline triclinic form. It is the bioactive constituent of the volatile oil of black seed. TQ exhibits anti-inflammatory, antioxidant and anti-neoplastic effects both in vitro and in vivo.

Application

Thymoquinone has been used as an inhibitor of Escherichia coli ATP synthase and cell growth. It is also used as a drug against lower limb ischemia-reperfusion injury of the sciatic nerve and femoral muscle.
Thymoquinone is employed in preclinical studies as a neuroprotective and anticonvulsant agent.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

219.2 °F - closed cup

Flash Point(C)

104 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Hala Gali-Muhtasib et al.
The international journal of biochemistry & cell biology, 38(8), 1249-1253 (2005-11-30)
There has been growing interest in naturally occurring compounds with anti-cancer potential. Black seed is one of the most extensively studied plants. This annual herb grows in countries bordering the Mediterranean Sea and India. Thymoquinone (TQ) is the bioactive constituent
Sanjar Alam et al.
International journal of nanomedicine, 7, 5705-5718 (2012-11-28)
Chitosan (CS) nanoparticles of thymoquinone (TQ) were prepared by the ionic gelation method and are characterized on the basis of surface morphology, in vitro or ex vivo release, dynamic light scattering, and X-ray diffractometry (XRD) studies. Dynamic laser light scattering
Thymoquinone: A promising anti-cancer drug from natural sources
Gali-Muhtasib H
The International Journal of Biochemistry & Cell Biology, 38, 1249-1253 (2006)
Chern Chiuh Woo et al.
Biochemical pharmacology, 83(4), 443-451 (2011-10-19)
Thymoquinone is an active ingredient isolated from Nigella sativa and has been investigated for its anti-oxidant, anti-inflammatory and anticancer activities in both in vitro and in vivo models since its first extraction in 1960s. Its anti-oxidant/anti-inflammatory effect has been reported
Thymoquinone Inhibits Escherichia coli ATP Synthase and Cell Growth
Ahmad Z
PLoS ONE, 10 (2015)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service