Skip to Content
Merck

Skip To

COLLD-RO

Roche

Collagenase D

from Clostridium histolyticum

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Pack SizeSKUAvailabilityPrice

About This Item

UNSPSC Code:
12352204
EC Number:
NACRES:
NA.54
Biological source:
Clostridium histolyticum
Concentration:
(working concentration: 0.5 - 2.5 mg/ml)
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


biological source

Clostridium histolyticum

sterility

non-sterile

form

lyophilized

collagenase activity

>0.15 U/mg lyophilizate

packaging

pkg of 100 mg (11088858001), pkg of 2.5 g (11088882001), pkg of 500 mg (11088866001)

manufacturer/tradename

Roche

concentration

(working concentration: 0.5 - 2.5 mg/ml)

technique(s)

tissue processing: suitable

color

light brown

optimum pH

6.0-8.0

solubility

water: soluble

NCBI accession no.

application(s)

cell analysis

foreign activity

Clostripain 0.060 U/mg, Proteases 8 U/mg ( Azocoll), Trypsin <0.2 U/mg (with BAEE)

storage temp.

2-8°C

General description

Collagenase D is prepared from Clostridium histolyticum cultures by filtration, ammonium sulfate precipitation, dialysis, and lyophilization.

Application

Collagenase from C. histolyticum is widely used for the disaggregation of many types of tissues (e.g., lung, heart, muscle, bone, adipose tissue, liver, kidney, cartilage, mammary gland, placentae, blood vessels, brain, tumors) and for the preparation of single cell suspensions for the establishment of primary cell culture systems. Collagenase D is recommended when functionality and integrity of cell-surface proteins are important.
Clostridium collagenase from Roche has been used to prepare cells from many types of tissue, such as hepatocytes, adipocytes, pancreatic islets, epithelial cells, muscle cells, endothelial cells, etc.[1][2][3][4] However, suitability of each lot of the enzyme for disruption of a particular tissue should be determined empirically.

Biochem/physiol Actions

Collagenase degrades native collagen. Clostripain, trypsin-like enzymes, and neutral proteases degrade other proteins as well.

Features and Benefits

Contents
Lyophilizate, nonsterile

Physical form

Lyophilizate, nonsterile.

Preparation Note

Activator: Ca2+
Working concentration: 0.5 - 2.5mg/ml
Storage conditions (working solution): -15 to -25°C
Roche recommends reconstituting only the amount of lyophilizate needed for immediate use. The reconstituted solution can be stored at -15 to -25°C for up to one week. Avoid repeated freezing and thawing since activity decreases after reconstitution.

Inhibitors:

Collagenase inhibitors: EDTA, EGTA, Cys, His, DTT, 2-mercaptoethanol
Collagenase is not inhibited by serum.
Clostripain inhibitors: TLCK
Trypsin inhibitors: aprotinin, trypsin inhibitor (egg white, soybean)

Enzyme activity:
Collagenase activity: >0.15 U/mg (according to Wünsch) (+25°C, 4-phenyl-azobenzyl-oxycarbonyl-Pro-Leu-Gly-Pro-D-Arg as the substrate)
Contaminating enzyme activities: trypsin, clostripain, and total proteolytic activity
Collagenase D has a normal to high collagenase activity and very low tryptic activity (usually <0.2 units/mg, BAEE as substrate).
Keep protected from light
Reconstitution in any balanced salt solution (e.g., HBSS)

Other Notes

Collagenase from Roche is assayed in Wünsch units (1 μmol of product formed per minute at +25 °C with Wünsch substrate).
Frequently, collagenase activities are given in Mandl units (1 μmol leucine liberated from collagen in 5 hours at +37 °C).
Unfortunately, there is no consistent conversion factor between the two units of activity, since the Mandl unit depends, in part, on the concentration of contaminating proteases in the collagenase preparation, an indefinable variable. A purer collagenase preparation would actually give a lower specific activity in Mandl units than a crude preparation. Clostridium preparations typically give conversion factors of approximately 1:1800 (e.g., a particular lot of Clostridium collagenase contained approximately 0.15 Wünsch U/mg and 250 Mandl U/mg).
For life science research only. Not for use in diagnostic procedures.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
COLLH-ROC9407COLLDISP-RO
biological source

Clostridium histolyticum

biological source

Clostridium histolyticum

biological source

Clostridium histolyticum

biological source

enzyme from Bacillus polymyxa ( Dispase ), enzyme from bacterial ( Collagenase from Vibrio alginolyticus)

technique(s)

tissue processing: suitable

technique(s)

tissue processing: suitable

technique(s)

cell culture | mammalian: suitable

technique(s)

tissue processing: suitable

application(s)

cell analysis

application(s)

life science and biopharma

application(s)

-

application(s)

life science and biopharma
sample preparation

concentration

(working concentration: 0.5 - 2.5 mg/ml)

concentration

~1 mg/mL (Isolation of rat hepatocytes), ~2 mg/mL (Isolation of adipocytes)

concentration

-

concentration

1 mg/mL (Collagenase: 0.1U/mL, Dispase: 0.8U/mL)

form

lyophilized

form

lyophilized

form

powder

form

lyophilized

solubility

water: soluble

solubility

water: soluble

solubility

-

solubility

water: soluble


pictograms

Exclamation markHealth hazard

signalword

Danger

Hazard Classifications

Eye Irrit. 2 - Resp. Sens. 1 - Skin Irrit. 2 - STOT SE 3

target_organs

Respiratory system

Storage Class

11 - Combustible Solids

wgk

WGK 1

flash_point_f

does not flash

flash_point_c

does not flash



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Charles J Benck et al.
Journal of visualized experiments : JoVE, (107)(107), e53638-e53638 (2016-02-11)
Dissociating murine skin into a single cell suspension is essential for downstream cellular analysis such as the characterization of infiltrating immune cells in rodent models of skin inflammation. Here, we describe a protocol for the digestion of mouse skin in
Chibuzor Uchea et al.
Toxicology research, 4(2), 494-507 (2015-04-22)
Prediction of xenobiotic fate in fish is important for the regulatory assessment of chemicals under current legislation. Trout hepatocyte spheroids are a promising
Qiuhua Yang et al.
British journal of pharmacology, 179(8), 1661-1678 (2021-11-20)
Excess nutrient-induced endothelial cell inflammation is a hallmark of high fat diet (HFD)-induced metabolic syndrome. Pharmacological activation of the protein kinase AMP-activated α1 (PRKAA1) also known as AMPKα1, shows its beneficial effects in many studies of cardiometabolic disorders. However, AMPKα1



Global Trade Item Number

SKUGTIN
1108885800104061838250612
1108888200104061838250636
1108886600104061838250629