Skip to Content
Merck
All Photos(1)

Documents

COLLDISP-RO

Roche

Collagenase/Dispase®

lyophilized, non-sterile, optimum pH 7.0-8.0

Synonym(s):

collagenase

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352204

biological source

enzyme from Bacillus polymyxa ( Dispase )
enzyme from bacterial ( Collagenase from Vibrio alginolyticus)

Quality Level

sterility

non-sterile

form

lyophilized

collagenase activity

>0.1 U/mg lyophilizate, 25 °C (4-phenyl-azobenzyl-oxycarbonyl-Pro-Leu-Gly-Pro-D-Arg as substrate)

specific activity

>0.8 U/mg lyophilizate, pH 7.5, 37 °C (casein as substrate, Dispase)

packaging

pkg of 100 mg (10269638001)
pkg of 500 mg (11097113001)

manufacturer/tradename

Roche

concentration

1 mg/mL (Collagenase: 0.1U/mL, Dispase: 0.8U/mL)

technique(s)

tissue processing: suitable

color

light yellow to brown

optimum pH

7.0-8.0

solubility

water: soluble

UniProt accession no.

application(s)

life science and biopharma
sample preparation

foreign activity

proteolytic activities <0.1 U/mg (hemoglobin as the substrate)

storage temp.

2-8°C

General description

Dispase is a neutral, rapid-acting, gentle protease that separates intact epidermis from dermis. It also separates cultured intact epithelial cells from the substratum. It cleaves the basement membrane zone and leaves the epithelial cells intact. Collagenase is a protease which cleaves the triple-helical protein called collagen. There are three types of tissue collagenases, and these belong to the matrix metalloproteinases (MMP) family. Collagenase obtained from Clostridium histolyticum has a very strong activity, as it digests collagen from both ends, at temperatures as low as 4-10 °C.

Specificity

Collagenase degrades native collagen. Dispase® is a nonspecific protease.

Application

Collagenase/Dispase® is used for the preparation of cells from a wide variety of different tissues and organs.

Components

EC 3.4.24.4 & 3.4.24.3

Unit Definition

One unit is the enzyme activity, which liberates under assay conditions within 1 minute Folin-positive amino acids and peptides corresponding to 1 μmol tyrosine.

Preparation Note

Activator: Ca2+

Working concentration: Approximately 1mg/mL (Collagenase: 0.1U/mL, Dispase: 0.8U/mL)

Enzyme activity:
Collagenase: 0.1 Wünsch U/mg lyophilizate (+25°C, 4-phenyl-azobenzyl-oxycarbonyl-Pro-Leu-Gly-Pro-D-Arg as the substrate)
Dispase®: 0.8 Wünsch U/mg lyophilizate (+37°C, casein as substrate, pH 7.5)
Contaminating proteolytic activities: <0.1U/mg lyophilizate (hemoglobin as the substrate)

Working solution: Preparation of working solution Dissolve enzyme in water to a final concentration of 100mg/mL. Dilute with PBS (Ca2+/Mg2+-free) to a final concentration of 1mg/mL and filter through a sterile 0.2μm membrane filter.
Enzyme concentration of the sterile solution: Collagenase, 0.1U/mL PBS; Dispase®, 0.8U/mL PBS.

Inhibitors:
Collagenase inhibitors: EDTA, EGTA
Collagenase/Dispase is not inhibited by serum.

Storage conditions (working solution): -15 to -25°C
The reconstituted solution is stable at -15 to -25°C.
Note: Store in appropriate aliquots and avoid repeated freezing and thawing!

Reconstitution

Reconstitution in water (100mg/mL). Further dilution with PBS (phosphate buffered saline) (Ca2+/Mg2+-free).

Storage and Stability

Store dry

Other Notes

Collagenase from Vibrio alginolyticus, Dispase® from Bacillus polymyxa
For life science research only. Not for use in diagnostic procedures.

Legal Information

Dispase is a registered trademark of Godo Shusei Co., Ltd.

Pictograms

Exclamation markHealth hazard

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Resp. Sens. 1 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

does not flash

Flash Point(C)

does not flash


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Elisa Seria et al.
BMC molecular and cell biology, 22(1), 28-28 (2021-05-12)
Chronic leg ulcerations are associated with Haemoglobin disorders, Type2 Diabetes Mellitus, and long-term venous insufficiency, where poor perfusion and altered metabolism develop into a chronic inflammation that impairs wound closure. Skin equivalent organotypic cultures can be engineered in vitro to
Sara Y Brucker et al.
Disease models & mechanisms, 15(5) (2022-04-09)
The uterus is responsible for the nourishment and mechanical protection of the developing embryo and fetus and is an essential part in mammalian reproduction. Mayer-Rokitansky-Küster-Hauser (MRKH) syndrome is characterized by agenesis of the uterus and upper part of the vagina
Winfried Neuhaus et al.
PloS one, 10(8), e0136221-e0136221 (2015-08-15)
Antenatal steroid treatment decreases morbidity and mortality in premature infants through the maturation of lung tissue, which enables sufficient breathing performance. However, clinical and animal studies have shown that repeated doses of glucocorticoids such as dexamethasone and betamethasone lead to
E Rotgers et al.
Andrology, 3(5), 947-955 (2015-08-11)
Accurate analysis and quantification of different testicular cell populations are of central importance in studies of male reproductive biology. The traditional histomorphometric and immunohistochemical methods remain the gold standard in studying the complex dynamics of the testicular tissue. Through past
Jack A Supple et al.
Current biology : CB, 30(4), 645-656 (2020-01-21)
Akin to all damselflies, Calopteryx (family Calopterygidae), commonly known as jewel wings or demoiselles, possess dichoptic (separated) eyes with overlapping visual fields of view. In contrast, many dragonfly species possess holoptic (dorsally fused) eyes with limited binocular overlap. We have

Articles

Organoid culture products to generate tissue and stem cell derived 3D brain, intestinal, gut, lung and cancer tumor organoid models.

Organoid culture products to generate tissue and stem cell derived 3D brain, intestinal, gut, lung and cancer tumor organoid models.

Organoid culture products to generate tissue and stem cell derived 3D brain, intestinal, gut, lung and cancer tumor organoid models.

Organoid culture products to generate tissue and stem cell derived 3D brain, intestinal, gut, lung and cancer tumor organoid models.

Related Content

Collagenase Guide.Collagenases, enzymes that break down the native collagen that holds animal tissues together, are made by a variety of microorganisms and by many different animal cells.

Collagenase Guide.Collagenases, enzymes that break down the native collagen that holds animal tissues together, are made by a variety of microorganisms and by many different animal cells.

Collagenase Guide.Collagenases, enzymes that break down the native collagen that holds animal tissues together, are made by a variety of microorganisms and by many different animal cells.

Collagenase Guide.Collagenases, enzymes that break down the native collagen that holds animal tissues together, are made by a variety of microorganisms and by many different animal cells.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service