Skip to Content
Merck

Skip To

ACOA-RO

Roche

Acetyl-Coenzyme A

85% (Enzymatic and Absorbance), 2% (lithium)

Synonym(s):

Acetyl-Coenzyme A, acetyl-coa

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Pack SizeSKUAvailabilityPrice

About This Item

UNSPSC Code:
12352204
NACRES:
NA.54
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


description

Chemical Formula: C23H35N7O17P3SLi3

assay

85% (Enzymatic and Absorbance)

form

solid

mol wt

Mr 809.6 (acetyl-CoA), Mr 827.4 (acetyl-CoA-Li3)

packaging

pkg of 010 mg (10101893001), pkg of 200 mg (11585371001), pkg of 050 mg (10101907001)

manufacturer/tradename

Roche

concentration

2% (lithium)

storage temp.

2-8°C

General description

Acetyl-Coenzyme A, trilithium salt

Application

Acetyl-Coenzyme A is used to assay CAT enzyme activity in cell extracts using radioisotopes. CAT enzyme activity in cell extracts catalyzes the transfer of acetyl groups from acetylcoenzyme A to chloramphenicol. A number of assays have been developed to measure CAT activity in cell extracts.[1] Acetyl-Coenzyme A has also been used to determine citrate synthase activity.

Biochem/physiol Actions

Acetyl-Coenzyme A (Ac-CoA) is the end product of glycolysis and takes part in the Ac-CoA pathway, which is a metabolic pathway for carbon compounds. Ac-CoA is important in cholesterol synthesis.[2][3] It also takes part in fatty acid biosynthesis and catabolism of polyamines like spermine and spermidine. A low level of Ac-CoA leads to a loss in glial cell and neuronal function. Ketone bodies and triglycerides give rise to Ac-CoA on hydrolysis and this indirectly leads to increased histone acetylation.[4][5][6]

Preparation Note

Working solution: Optimal solvent: water or aqueous solution with a weak acidic pH (4 to 5).
Storage conditions (working solution): -15 to -25 °C
50 mg/ml in phosphate buffer, pH 7, is stable for 3 weeks at -15 to -25 °C. Unfrozen solutions should be immediately used.

Other Notes

For life science research only. Not for use in diagnostic procedures.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
A2056P5397870724P
assay

85% (Enzymatic and Absorbance)

assay

≥93% (HPLC)

assay

≥85%

assay

>99% (TLC)

form

solid

form

powder

form

powder

form

powder

storage temp.

2-8°C

storage temp.

−20°C

storage temp.

−20°C

storage temp.

−20°C

manufacturer/tradename

Roche

manufacturer/tradename

-

manufacturer/tradename

-

manufacturer/tradename

Avanti Research - A Croda Brand 870724P

description

Chemical Formula: C23H35N7O17P3SLi3

description

-

description

-

description

-

mol wt

Mr 809.6 (acetyl-CoA), Mr 827.4 (acetyl-CoA-Li3)

mol wt

-

mol wt

-

mol wt

-


Storage Class

11 - Combustible Solids

wgk

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Biochemistry null
Giosalba Burgio et al.
Biochimica et biophysica acta, 1799(10-12), 671-680 (2010-05-25)
The eukaryotic genome is a highly organized nucleoprotein structure comprising of DNA, histones, non-histone proteins, and RNAs, referred to as chromatin. The chromatin exists as a dynamic entity, shuttling between the open and closed forms at specific nuclear regions and
Morten Scheibye-Knudsen et al.
Cell metabolism, 20(5), 840-855 (2014-12-03)
Cockayne syndrome (CS) is an accelerated aging disorder characterized by progressive neurodegeneration caused by mutations in genes encoding the DNA repair proteins CS group A or B (CSA or CSB). Since dietary interventions can alter neurodegenerative processes, Csb(m/m) mice were



Global Trade Item Number

SKUGTIN
1010189300104061838665454
1010190700104061838250384
1158537100104061838251084