Skip to Content
Merck
All Photos(2)

Key Documents

P5397

Sigma-Aldrich

n-Propionyl coenzyme A lithium salt

≥85%

Synonym(s):

n-Propionyl CoA lithium salt

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C24H40N7O17P3S
CAS Number:
Molecular Weight:
823.60
UNSPSC Code:
41106305
PubChem Substance ID:
NACRES:
NA.51

biological source

yeast

Assay

≥85%

form

powder

solubility

H2O: soluble-50 mg/mL, clear, colorless

storage temp.

−20°C

SMILES string

[Li].[P](=O)(O[P](=O)(OCC([C@@H](O)C(=O)NCCC(=O)NCCSC(=O)CC)(C)C)O)(OC[C@H]1O[C@H]([C@@H]([C@@H]1O[P](=O)(O)O)O)[n]2c3ncnc(c3nc2)N)O

InChI

1S/C24H40N7O17P3S.Li/c1-4-15(33)52-8-7-26-14(32)5-6-27-22(36)19(35)24(2,3)10-45-51(42,43)48-50(40,41)44-9-13-18(47-49(37,38)39)17(34)23(46-13)31-12-30-16-20(25)28-11-29-21(16)31;/h11-13,17-19,23,34-35H,4-10H2,1-3H3,(H,26,32)(H,27,36)(H,40,41)(H,42,43)(H2,

InChI key

CXNLEKKSVXVZAF-IEQRABFGSA-N

General description

Propionyl-CoA is obtained as an end product of isoleucine, valine and methionine catabolism. It is an essential component for the methylaspartate cycle. Branched-chain amino acids and cholesterol also give rise to propionyl-CoA. Propionyl coenzyme A (CoA) is the coenzyme A derivative of propionic acid. Propionyl CoA is formed during the β-oxidation of odd-chain fatty acids.

Biochem/physiol Actions

Coenzyme A functions as an acyl group carrier, acetyl-CoA. Propionyl coenzyme A (Propionyl-CoA) may be used to characterize and study propionyl-coenzyme A carboxylase complexes found in bacteria. Propionyl-CoA may be used to study the specificity and kinetics of methylisocitrate lyase(s).

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

TetR family transcriptional regulator PccD negatively controls propionyl coenzyme A assimilation in Saccharopolyspora erythraea
Xu Z, et al.
Journal of Bacteriology, 199(20), e00281-e00217 (2017)
Production of 3-Hydroxypropionic Acid via the Propionyl-CoA Pathway Using Recombinant Escherichia coli Strains
Luo H, et al.
Testing, 11(5), e0156286-e0156286 (2016)
Atanas V Demirev et al.
Journal of microbiology (Seoul, Korea), 49(3), 407-412 (2011-07-01)
Acyl-CoA carboxylases (ACC) are involved in important primary or secondary metabolic pathways such as fatty acid and/or polyketides synthesis. In the 62 kb fragment of pccB gene locus of Streptomyces toxytricini producing a pancreatic inhibitor lipstatin, 3 distinct subunit genes
Propionyl coenzyme A (propionyl-CoA) carboxylase in Haloferax mediterranei: indispensability for propionyl-CoA assimilation and impacts on global metabolism
Hou J, et al.
Applied and Environmental Microbiology, 81(2), 794-804 (2015)
M A Luttik et al.
Journal of bacteriology, 182(24), 7007-7013 (2000-11-28)
The Saccharomyces cerevisiae ICL1 gene encodes isocitrate lyase, an essential enzyme for growth on ethanol and acetate. Previous studies have demonstrated that the highly homologous ICL2 gene (YPR006c) is transcribed during the growth of wild-type cells on ethanol. However, even

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service