Skip to Content
Merck

Skip To

PHR2149

Phenylephrine Related Compound F

Pharmaceutical Secondary Standard; Certified Reference Material

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Pack SizeSKUAvailabilityPrice

About This Item

NACRES:
NA.24
UNSPSC Code:
41116107
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Segment

grade

certified reference material, pharmaceutical secondary standard

agency

traceable to USP 1533261

API family

phenylephrine

CofA

current certificate can be downloaded

packaging

pkg of 100 mg

application(s)

pharmaceutical small molecule

format

neat

storage temp.

2-8°C

General description

Pharmaceutical secondary standards for application in quality control provide pharma laboratories and manufacturers with a convenient and cost-effective alternative to the preparation of in-house working standards

Application

These Secondary Standards are qualified as Certified Reference Materials. These are suitable for use in several analytical applications including but not limited to pharma release testing, pharma method development for qualitative and quantitative analyses, food and beverage quality control testing, and other calibration requirements.

Analysis Note

These secondary standards offer multi-traceability to the USP and EP primary standards, where they are available.

Other Notes

This Certified Reference Material (CRM) is produced and certified in accordance with ISO 17034 and ISO/IEC 17025. All information regarding the use of this CRM can be found on the certificate of analysis.
To see an example of a Certificate of Analysis for this material enter LRAB9879 in the Documents slot below. This is an example certificate only and may not be the lot that you receive.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
PHR1150PHR1755PHR2423
application(s)

pharmaceutical small molecule

application(s)

pharmaceutical (small molecule)

application(s)

pharmaceutical (small molecule)

application(s)

pharmaceutical

format

neat

format

neat

format

-

format

-

Quality Level

300

Quality Level

300

Quality Level

300

Quality Level

300

grade

certified reference material, pharmaceutical secondary standard

grade

certified reference material, pharmaceutical secondary standard

grade

certified reference material, pharmaceutical secondary standard

grade

certified reference material, pharmaceutical secondary standard

storage temp.

2-8°C

storage temp.

2-30°C

storage temp.

-10 to -25°C

storage temp.

2-30°C

packaging

pkg of 100 mg

packaging

-

packaging

pkg of 30 mg

packaging

pkg of 100 mg


Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable



Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters