Select a Size
| Size/SKU | Availability | Price |
|---|
About This Item
biological source
rabbit
Quality Segment
conjugate
unconjugated
antibody form
purified antibody
antibody product type
primary antibodies
clone
polyclonal
mol wt
calculated mol wt 69.76 kDa, observed mol wt ~105 kDa
purified by
affinity chromatography
species reactivity
human
packaging
antibody small pack of 100 μg
technique(s)
immunohistochemistry: suitable, western blot: suitable
isotype
IgGκ
epitope sequence
C-terminal
Protein ID accession no.
UniProt accession no.
storage temp.
2-8°C
target post-translational modification
unmodified
Gene Information
human ... UMOD(7369)
General description
Immunogen
Application
Evaluated by Western Blotting in Human kidney tissue lysate.
Western Blotting Analysis: A 1:1,000 dilution of this antibody detected Uromodulin/THP in Human kidney tissue lysate.
Tested applications
Immunohistochemistry (Paraffin) Analysis: A 1:1,000 dilution from a representative lot detected Uromodulin/THP in human kidney tissue sections.
Note: Actual optimal working dilutions must be determined by end user as specimens, and experimental conditions may vary with the end user
Biochem/physiol Actions
Physical form
Preparation Note
Other Notes
Disclaimer
1 of 1
This Item | |||
|---|---|---|---|
| conjugate unconjugated | conjugate unconjugated | conjugate unconjugated | conjugate unconjugated |
| antibody form purified antibody | antibody form purified immunoglobulin | antibody form IgG fraction of antiserum | antibody form affinity isolated antibody |
| species reactivity human | species reactivity human | species reactivity human | species reactivity human |
| clone polyclonal | clone 3F10, monoclonal | clone polyclonal | clone polyclonal |
| biological source rabbit | biological source mouse | biological source mouse | biological source rabbit |
| technique(s) immunohistochemistry: suitable, western blot: suitable | technique(s) capture ELISA: suitable, immunoprecipitation (IP): suitable, western blot: 1-5 μg/mL | technique(s) immunohistochemistry (formalin-fixed, paraffin-embedded sections): suitable, indirect immunofluorescence: suitable, western blot: 1 μg/mL | technique(s) immunohistochemistry: 1:1000-1:2500 |
Still not finding the right product?
Try our Product Selector Tool to narrow your options
Storage Class
12 - Non Combustible Liquids
wgk
WGK 2
flash_point_f
Not applicable
flash_point_c
Not applicable
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.



