Skip to Content
Merck

Skip To

8.16019

(S)-(-)-Proline

for synthesis

Synonym(s):

(S)-(-)-Proline, Pro

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Pack SizeSKUAvailabilityPrice

About This Item

Empirical Formula (Hill Notation):
C5H9NO2
CAS Number:
Molecular Weight:
115.13
UNSPSC Code:
12352209
EC Index Number:
205-702-2
NACRES:
NA.22
MDL number:
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Segment

form

solid

potency

>5110 mg/kg LD50, oral (Rat)

pH

5-7 (20 °C, 100 g/L in H2O)

mp

220-222 °C (decomposition)

solubility

1500 g/L

density

1.35-1.38 g/cm3 at 25 °C

bulk density

500 kg/m3

storage temp.

2-30°C

SMILES string

[N+H2]1C(CCC1)C(=O)[O-]

InChI

1S/C5H9NO2/c7-5(8)4-2-1-3-6-4/h4,6H,1-3H2,(H,7,8)

InChI key

ONIBWKKTOPOVIA-UHFFFAOYSA-N

Analysis Note

Assay (HClO₄): ≥ 99.0 %
Spec. rotation [α²0/D (c=4 in water): -86.0 - -84.0 °
Identity (IR): passes test

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
1.074341.07430PHL83414
form

solid

form

solid

form

-

form

solid

storage temp.

2-30°C

storage temp.

2-25°C

storage temp.

2-30°C

storage temp.

-

Quality Level

200

Quality Level

100

Quality Level

500

Quality Level

-

solubility

1500 g/L

solubility

1500 g/L

solubility

1500 g/L

solubility

-

mp

220-222 °C (decomposition)

mp

220-222 °C (decomposition)

mp

220-222 °C (decomposition)

mp

228 °C (dec.) (lit.)

density

1.35-1.38 g/cm3 at 25 °C

density

1.35-1.38 g/cm3 at 25 °C

density

1.35-1.38 g/cm3 at 25 °C

density

-


Storage Class

13 - Non Combustible Solids

wgk

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable



Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library







Global Trade Item Number

SKUGTIN
816019002504022536423937
816019010004022536423944

Questions

Reviews

No rating value

Active Filters