Sign In to View Organizational & Contract Pricing.
Select a Size
Change View
| Pack Size | SKU | Availability | Price |
|---|
About This Item
UNSPSC Code:
12352204
EC Number:
232-619-9
NACRES:
NA.54
Pricing and availability is not currently available.
storage temp.
2-8°C
Quality Segment
Other Notes
Composition:
62301 Lipase from Aspergillus niger, 100 mg
62288 Lipase B Candida antarctica, recombinant from Aspergillus oryzae, 50 mg
62316 Lipase from Candida rugosa, 1000 mg
62298 Lipase from Mucor miehei, 100 mg
62309 Lipase from Pseudomonas cepacia, 100 mg
89601 Lipase from Pseudomonas fluorescens, 200 mg
62305 Lipase from Rhizopus arrhizus, 1000 mg
62310 Lipase from Rhizopus niveus, 1000 mg
L3126 Lipase from porcine pancreas, 1000 mg
62301 Lipase from Aspergillus niger, 100 mg
62288 Lipase B Candida antarctica, recombinant from Aspergillus oryzae, 50 mg
62316 Lipase from Candida rugosa, 1000 mg
62298 Lipase from Mucor miehei, 100 mg
62309 Lipase from Pseudomonas cepacia, 100 mg
89601 Lipase from Pseudomonas fluorescens, 200 mg
62305 Lipase from Rhizopus arrhizus, 1000 mg
62310 Lipase from Rhizopus niveus, 1000 mg
L3126 Lipase from porcine pancreas, 1000 mg
1 of 1
This Item | |||
|---|---|---|---|
| storage temp. 2-8°C | storage temp. −70°C | storage temp. −70°C | storage temp. 2-8°C |
| Quality Level 100 | Quality Level 200 | Quality Level 200 | Quality Level - |
signalword
Danger
hcodes
pcodes
Hazard Classifications
Resp. Sens. 1
Storage Class
10 - Combustible liquids
flash_point_f
Not applicable
flash_point_c
Not applicable
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Global Trade Item Number
| SKU | GTIN |
|---|---|
| 62327-1EA | 04061832754352 |




