Skip to Content
Merck

Skip To

72129

Neoeriocitrin

~95% (HPLC)

Synonym(s):

(S)-3′,4′,5,7-Tetrahydroxyflavanone-7-[2-O-(α-L-rhamnopyranosyl)-β-D-glucopyranoside], Eriodictyol 7-neohesperidoside

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Pack SizeSKUAvailabilityPrice

About This Item

Empirical Formula (Hill Notation):
C27H32O15
CAS Number:
Molecular Weight:
596.53
UNSPSC Code:
85151701
NACRES:
NA.47
PubChem Substance ID:
EC Number:
236-215-3
Beilstein/REAXYS Number:
1696764
MDL number:
Assay:
~95% (HPLC)
Form:
powder
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


assay

~95% (HPLC)

form

powder

SMILES string

C[C@@H]1O[C@@H](O[C@@H]2[C@@H](O)[C@H](O)[C@@H](CO)O[C@H]2Oc3cc(O)c4C(=O)C[C@H](Oc4c3)c5ccc(O)c(O)c5)[C@H](O)[C@H](O)[C@H]1O

InChI

1S/C27H32O15/c1-9-20(33)22(35)24(37)26(38-9)42-25-23(36)21(34)18(8-28)41-27(25)39-11-5-14(31)19-15(32)7-16(40-17(19)6-11)10-2-3-12(29)13(30)4-10/h2-6,9,16,18,20-31,33-37H,7-8H2,1H3/t9-,16-,18+,20-,21+,22+,23-,24+,25+,26-,27+/m0/s1

InChI key

OBKKEZLIABHSGY-DOYQYKRZSA-N

Biochem/physiol Actions

Neoeriocitrin was one of several flavonoids that completely blocked the okadaic acid-induced inhibition of autophagy in cultured mouse hepatocytes. It is suggested as tool in the study of intracellular protein phosphorylation and as a potential protectant against pathological hyperphosphorylations.


Still not finding the right product?

Explore all of our products under Neoeriocitrin


Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library







Global Trade Item Number

SKUGTIN
72129-5MG04061832404981
72129-1MG04061832856216

Questions

Reviews

No rating value

Active Filters