Skip to Content
Merck

Skip To

1.00993

2-Propanol

greener alternative

≥99.9% (GC), suitable for UV/Vis spectroscopy, Uvasol®

Synonym(s):

sec-Propyl alcohol, IPA, Isopropanol, Isopropyl alcohol

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Pack SizeSKUAvailabilityPrice

About This Item

Linear Formula:
(CH3)2CHOH
CAS Number:
Molecular Weight:
60.10
NACRES:
NA.03
EC Index Number:
200-661-7
UNSPSC Code:
12191502
MDL number:
Beilstein/REAXYS Number:
635639
Assay:
≥99.9% (GC)
Technique(s):
UV/Vis spectroscopy: suitable
Bp:
82 °C (lit.)
Vapor pressure:
33 mmHg ( 20 °C), 43 hPa ( 20 °C), 44 mmHg ( 25 °C)
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Product Name

2-Propanol, for spectroscopy Uvasol®

vapor density

2.1 (vs air)

Quality Segment

vapor pressure

33 mmHg ( 20 °C), 43 hPa ( 20 °C), 44 mmHg ( 25 °C)

assay

≥99.9% (GC)

form

liquid

autoignition temp.

750 °F

potency

5045 mg/kg LD50, oral (Rat), 12800 mg/kg LD50, skin (Rabbit)

expl. lim.

2.0-12.7 %, 93 °C

greener alternative product characteristics

Safer Solvents and Auxiliaries
Learn more about the Principles of Green Chemistry.

sustainability

Greener Alternative Product

technique(s)

UV/Vis spectroscopy: suitable

impurities

≤0.0001 meq/g Acidity, ≤0.0001 meq/g Alkalinity, ≤0.05% Water

evapn. residue

≤0.0002%

color

APHA: ≤10

transmittance

210 nm, ≥30%, 220 nm, ≥65%, 230 nm, ≥80%, 240 nm, ≥90%, 250 nm, ≥95%, 260 nm, ≥98%

refractive index

n20/D 1.377 (lit.)

pH

7 (20 °C)

bp

82 °C (lit.)

mp

−89.5 °C (lit.)

transition temp

flash point 12 °C

density

0.785 g/mL at 25 °C (lit.)

UV absorption

λ: 210 nm Amax: ≤0.52, λ: 220 nm Amax: ≤0.19, λ: 230 nm Amax: ≤0.10, λ: 240 nm Amax: ≤0.05, λ: 250 nm Amax: ≤0.02, λ: 260 nm Amax: ≤0.01

greener alternative category

storage temp.

2-25°C

SMILES string

CC(C)O

InChI

1S/C3H8O/c1-3(2)4/h3-4H,1-2H3

InChI key

KFZMGEQAYNKOFK-UHFFFAOYSA-N

General description

Accurate analytic results in UV/VIS and infrared spectroscopy depend on the use of very pure solvents for sample preparation. The Uvasol<TMSYMBOL></TMSYMBOL> solvents range has been specially designed for spectroscopy and other applications requiring solvents of the highest spectral purity. The refinement process allows a greater degree of security in applications and avoids misinterpretation of analytical results caused by traces of UV, IR and fluorescence contamination. Uvasol<TMSYMBOL></TMSYMBOL> solvents offer best UV transmittance. In all specifications the minimum transmittance for 5 typical wavelengths are identified.

Features and Benefits

We are committed to bringing you Greener Alternative Products, which adhere to one or more of The 12 Principles of Greener Chemistry. This solvent is safe to use and thus aligns with "Safer Solvents and Auxiliaries". Click here for more information.

Other Notes

Explore various lab safety accessories and equipment for safe handling of solvents to increase your safety level from the first usage.

Legal Information

UVASOL is a registered trademark of Merck KGaA, Darmstadt, Germany

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
1.010401.00998109634
Quality Level

100

Quality Level

100

Quality Level

100

Quality Level

300

technique(s)

UV/Vis spectroscopy: suitable

technique(s)

HPLC: suitable

technique(s)

gas chromatography (GC): suitable

technique(s)

-

assay

≥99.9% (GC)

assay

≥99.9% (GC)

assay

≥99.8% (GC)

assay

≥99.8% (GC)

form

liquid

form

liquid

form

liquid

form

liquid

density

0.785 g/mL at 25 °C (lit.)

density

0.785 g/mL at 25 °C (lit.)

density

0.785 g/mL at 25 °C (lit.)

density

0.785 g/mL at 25 °C (lit.)

bp

82 °C (lit.)

bp

82 °C (lit.)

bp

82 °C (lit.)

bp

82 °C (lit.)


Still not finding the right product?

Explore all of our products under 2-Propanol


pictograms

FlameExclamation mark

signalword

Danger

Hazard Classifications

Eye Irrit. 2 - Flam. Liq. 2 - STOT SE 3

target_organs

Respiratory system

Storage Class

3 - Flammable liquids

wgk

WGK 1

flash_point_f

53.6 °F - closed cup

flash_point_c

12.0 °C - closed cup



Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library





Global Trade Item Number

SKUGTIN
100993250004022536013855
100993100004022536013831