Skip to Content
Merck

Skip To

1485205

USP

Oxycodone hydrochloride

United States Pharmacopeia (USP) Reference Standard

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Pack SizeSKUAvailabilityPrice

About This Item

Empirical Formula (Hill Notation):
C18H21NO4 · HCl
CAS Number:
Molecular Weight:
351.82
NACRES:
NA.24
PubChem Substance ID:
UNSPSC Code:
41116107
MDL number:
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


grade

pharmaceutical primary standard

API family

oxycodone

manufacturer/tradename

USP

drug control

USDEA Schedule I; regulated under CDSA - not available from Sigma-Aldrich Canada; estupefaciente (Spain); Decreto Lei 15/93: Tabela IA (Portugal), kontrollierte Droge in Deutschland

application(s)

pharmaceutical (small molecule)

format

neat

SMILES string

Cl.[H][C@@]12Oc3c(OC)ccc4C[C@H]5N(C)CC[C@@]1(c34)[C@@]5(O)CCC2=O

InChI

1S/C18H21NO4.ClH/c1-19-8-7-17-14-10-3-4-12(22-2)15(14)23-16(17)11(20)5-6-18(17,21)13(19)9-10;/h3-4,13,16,21H,5-9H2,1-2H3;1H/t13-,16+,17+,18-;/m1./s1

InChI key

MUZQPDBAOYKNLO-RKXJKUSZSA-N

Gene Information

human ... OPRM1(4988)

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including MSDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia.
For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Oxycodone hydrochloride USP reference standard, intended for use in specified quality tests and assays as specified in the USP compendia. Also, for use with USP monographs such as:
  • Oxycodone Hydrochloride Tablets

Biochem/physiol Actions

μ and κ opioid receptor agonist.

Analysis Note

These products are for test and assay use only. They are not meant for administration to humans or animals and cannot be used to diagnose, treat, or cure diseases of any kind.  ​

Other Notes

Sales restrictions may apply.

Legal Information

German
Dieses Produkt fällt unter das Betäubungsmittelgesetz (BtMG). Für eine Bestellung dieses Produktes ist eine Erlaubnis nach § 3 BtMG zwingend erforderlich, es sei denn, es greift eine Ausnahme von der Erlaubnispflicht nach § 4 oder § 26 BtMG.

English
This product is subject to the German Narcotics Act. A permit under Section 3 of the German Narcotics Act is mandatory for ordering this product unless an exemption from the permit requirement under Section 4 or Section 26 of the German Narcotics Act applies.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
O1378BP10681323000
manufacturer/tradename

USP

manufacturer/tradename

-

manufacturer/tradename

BP

manufacturer/tradename

USP

grade

pharmaceutical primary standard

grade

analytical standard

grade

pharmaceutical primary standard

grade

pharmaceutical primary standard

application(s)

pharmaceutical (small molecule)

application(s)

forensics and toxicology
pharmaceutical (small molecule)
veterinary

application(s)

pharmaceutical
pharmaceutical small molecule

application(s)

pharmaceutical (small molecule)

format

neat

format

neat

format

neat

format

neat

API family

oxycodone

API family

-

API family

oxycodone

API family

hydromorphone

Gene Information

human ... OPRM1(4988)

Gene Information

human ... OPRM1(4988)

Gene Information

human ... OPRM1(4988)

Gene Information

human ... OPRM1(4988)


Still not finding the right product?

Explore all of our products under Oxycodone hydrochloride


pictograms

Exclamation mark

signalword

Warning

hcodes

Hazard Classifications

Acute Tox. 4 Oral

Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Global Trade Item Number

SKUGTIN
1485205-1G04061833860588

Questions

Reviews

No rating value

Active Filters