Sign In to View Organizational & Contract Pricing.
Select a Size
Change View
| Size/SKU | Availability | Price |
|---|
About This Item
Biological source:
mouse
Form:
lyophilized powder
Shipped in:
dry ice
Pricing and availability is not currently available.
biological source
mouse
Quality Segment
form
lyophilized powder
technique(s)
cell culture | embryo: suitable
shipped in
dry ice
storage temp.
−20°C
Application
Complement sera from mouse has been used for incubation of zona-free embryos prior to microscopic examination.
Physical form
Lyophilized powder from indicated amount of serum
Preparation Note
Reconstitute in the indicated amount of ice cold deionized water. Store at -70 °C after reconstitution. Avoid repeated freezing and thawing.
Analysis Note
Hemolytic titer (CH50units per ml) is determined by the method of Kabat and Mayer. Actual titer given on the label.
1 of 1
This Item | |||
|---|---|---|---|
| form lyophilized powder | form lyophilized powder | form lyophilized powder | form liquid |
| technique(s) cell culture | embryo: suitable | technique(s) cell culture | embryo: suitable | technique(s) cell culture | embryo: suitable, cell culture | mammalian: suitable | technique(s) cell culture | embryo: suitable |
| biological source mouse | biological source guinea pig | biological source rabbit | biological source - |
| shipped in dry ice | shipped in dry ice | shipped in dry ice | shipped in ambient |
| storage temp. −20°C | storage temp. −20°C | storage temp. −20°C | storage temp. 2-8°C |
| Quality Level 200 | Quality Level 200 | Quality Level 200 | Quality Level 300 |
wgk
WGK 3
Storage Class
11 - Combustible Solids
flash_point_f
Not applicable
flash_point_c
Not applicable
ppe
Eyeshields, Gloves, type N95 (US)
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.



