Merck
Skip To
C6807
Nunc® Lab-Tek® II Chamber Slide system
wells 4, well area 1.7 cm2, sterile
Synonym(s):
cell culture chamber, chamber slides, microscope slides, microscopy slides, Nunc 154526
Sign In to View Organizational & Contract Pricing.
Select a Size
Change View
Pack SizeSKUAvailabilityPrice
About This Item
NACRES:
NB.22
UNSPSC Code:
41122600
Material:
colorless , polystyrene chamber
Sterility:
sterile
Packaging:
pack of 16 ea, case of 96 ea
Wells:
4
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist
material
colorless , polystyrene chamber
sterility
sterile
feature
CE marked
packaging
pack of 16 ea, case of 96 ea
manufacturer/tradename
Nunc 154526
well area
1.7 cm2
wells
4
working volume
0.5-1.0 mL
suitability
suitable for (cell culture; attachment and growth of cells)
General description
Grow, stain, and examine cells directly on the Nunc® Lab-Tek® II chamber slide which consists of a removable polystyrene media chamber attached to a standard glass slide. Slide surface is specially treated for adherent cell culture. Available in 1-, 2-, 4-, and 8-well formats.
Features and Benefits
  • Slide surface is specially treated with a unique RS wash to ensure consistent cell growth.
  • Tabbed polystyrene lid allows gas exchange with minimal evaporation.
  • Removal tool separates media chamber from glass slide.
  • Biocompatible adhesive hydrophobic well border is printed on slide.
  • Slide features a white frosted writing area.
  • Includes: Chamber removal tool.
  • Sterile
  • Packed in convenient stackable incubation trays.
Legal Information
Chamber Slide is a trademark of Thermo Fisher Scientific or its subsidiaries
Lab-Tek is a registered trademark of Thermo Fisher Scientific or its subsidiaries
Nunc is a registered trademark of Thermo Fisher Scientific or its subsidiaries
Compare Similar Items
View Full Comparison
Show Differences
1 of 1
This Item
S6815S6565S6690
manufacturer/tradename
Nunc 154526
manufacturer/tradename
Nunc 154941
manufacturer/tradename
Nunc 154852
manufacturer/tradename
Nunc 154917
material
colorless , polystyrene chamber
material
colorless , polystyrene chamber
material
polystyrene chamber
material
colorless , polystyrene chamber
wells
4
wells
8
wells
-
wells
4
packaging
pack of 16 ea, case of 96 ea
packaging
pkg of carton of 16
packaging
pkg of carton of 16
packaging
pkg of carton of 16
working volume
0.5-1.0 mL
working volume
0.2-0.5 mL
working volume
1.0-2.0 mL
working volume
0.5-1.0 mL
well area
1.7 cm2
well area
0.7 cm2
well area
-
well area
1.7 cm2
Still not finding the right product?
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Visit the Document Library
Hao Sun et al.
Journal of cellular and molecular medicine, 20(7), 1234-1246 (2016-01-30)
Idiopathic pulmonary fibrosis (IPF) is a chronic fatal lung disease characterized by aberrant accumulation of fibroblast population and deposition of extra cellular matrix. Increasing evidence support that epithelial-mesenchymal transition (EMT) of alveolar epithelial cells is a critical process in the
Muzammel Haque et al.
Virology, 499, 156-164 (2016-09-24)
We have previously shown that the HSV-1 gK and UL20 proteins interact and function in virion envelopment, membrane fusion, and neuronal entry. Alignment of the predicted secondary structures of gKs encoded by BoHV-1, HSV-1, HSV-2, EHV-1 and VZV indicated a
Global Trade Item Number
SKUGTIN
C6807-1CS04061834373995
C6807-1PAK04061834374008