Skip to Content
Merck

Skip To

71505

Sodium phosphate monobasic dihydrate

BioUltra, Molecular Biology, ≥99.0% (T)

Synonym(s):

Sodium dihydrogen phosphate dihydrate

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Pack SizeSKUAvailabilityPrice

About This Item

Linear Formula:
NaH2PO4 · 2H2O
CAS Number:
Molecular Weight:
156.01
NACRES:
NA.21
PubChem Substance ID:
UNSPSC Code:
12161700
EC Number:
231-449-2
MDL number:
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


grade

Molecular Biology

Quality Level

product line

BioUltra

assay

≥99.0% (T)

form

powder or crystals

impurities

DNases, none detected, RNases, none detected, insoluble matter, passes filter test, phosphatases, none detected, proteases, none detected

pH

4.0-4.5 (25 °C, 1 M in H2O)

solubility

H2O: 1 M at 20 °C, clear, colorless

anion traces

chloride (Cl-): ≤50 mg/kg, sulfate (SO42-): ≤50 mg/kg

cation traces

Al: ≤5 mg/kg, As: ≤0.2 mg/kg, Ba: ≤5 mg/kg, Bi: ≤5 mg/kg, Ca: ≤10 mg/kg, Cd: ≤5 mg/kg, Co: ≤5 mg/kg, Cr: ≤5 mg/kg, Cu: ≤5 mg/kg, Fe: ≤5 mg/kg, K: ≤200 mg/kg, Li: ≤5 mg/kg, Mg: ≤5 mg/kg, Mn: ≤5 mg/kg, Mo: ≤5 mg/kg, Ni: ≤5 mg/kg, Pb: ≤5 mg/kg, Sr: ≤5 mg/kg, Zn: ≤5 mg/kg

λ

1 M in H2O

UV absorption

λ: 260 nm Amax: 0.085, λ: 280 nm Amax: 0.070

application(s)

diagnostic assay manufacturing

SMILES string

O.O.[Na+].OP(O)([O-])=O

InChI

1S/Na.H3O4P.2H2O/c;1-5(2,3)4;;/h;(H3,1,2,3,4);2*1H2/q+1;;;/p-1

InChI key

VBJGJHBYWREJQD-UHFFFAOYSA-M

Application

Sodium phosphate monobasic dehydrate has been used in the preparation of the phosphate buffer solution and SCCP solution. It has also been used in the preparation of stop buffer.

Other Notes

Gel electrophoresis - running buffer component for denaturing gels containing formamide.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
71500042691.28238
assay

≥99.0% (T)

assay

≥99.0% (T)

assay

98-100.5% (calc. to the dried substance)

assay

-

grade

Molecular Biology

grade

-

grade

-

grade

-

Quality Level

200

Quality Level

300

Quality Level

200

Quality Level

500

product line

BioUltra

product line

-

product line

-

product line

EMPROVE® EXPERT

form

powder or crystals

form

crystals

form

powder or crystals

form

crystalline

application(s)

diagnostic assay manufacturing

application(s)

-

application(s)

-

application(s)

liquid formulation
parenterals
pharma/biopharma processes


Still not finding the right product?


Storage Class

13 - Non Combustible Solids

wgk

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



In vitro assay to determine SUMOylation sites on protein substrates
McManus F P, et al.
Nature Protocols, 11(2), 387-387 (2016)
3D FISH to analyse gene domain-specific chromatin re-modeling in human cancer cell lines
Kocanova S, et al.
Methods (2018)
Gold nanoparticle/nickel oxide/poly (pyrrole-N-propionic acid) hybrid multilayer film: Electrochemical study and its application in biosensing
Karazehir T, et al.
Express Polymer Letters, 11(6), 449-449 (2017)



Global Trade Item Number

SKUGTIN
PHR1742-200MG04061834802198
117188-25G04061831818383
71505-1KG04061832848839
71505-250G04061832848846

Questions

1–2 of 2 Questions  
  1. How can I determine the shelf life / expiration / retest date of this product?

    1 answer
    1. If this product has an expiration or retest date, it will be shown on the Certificate of Analysis (COA, CofA). If there is no retest or expiration date listed on the product's COA, we do not have suitable stability data to determine a shelf life. For these products, the only date on the COA will be the release date; a retest, expiration, or use-by-date will not be displayed.
      For all products, we recommend handling per defined conditions as printed in our product literature and website product descriptions. We recommend that products should be routinely inspected by customers to ensure they perform as expected.
      For products without retest or expiration dates, our standard warranty of 1 year from the date of shipment is applicable.
      For more information, please refer to the Product Dating Information document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/418/501/product-dating-information-06-25-mk.pdf

      Helpful?

  2. How is shipping temperature determined? And how is it related to the product storage temperature?

    1 answer
    1. Products may be shipped at a different temperature than the recommended long-term storage temperature. If the product quality is sensitive to short-term exposure to conditions other than the recommended long-term storage, it will be shipped on wet or dry-ice. If the product quality is NOT affected by short-term exposure to conditions other than the recommended long-term storage, it will be shipped at ambient temperature. As shipping routes are configured for minimum transit times, shipping at ambient temperature helps control shipping costs for our customers. For more information, please refer to the Storage and Transport Conditions document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/316/622/storage-transport-conditions-mk.pdf

      Helpful?

Reviews

No rating value

Active Filters