Skip to Content
Merck

Skip To

RTC000102

o-Xylene

Pharmaceutical Secondary Standard; Certified Reference Material

Synonym(s):

o-Xylene, 1,2-Dimethylbenzene

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice

About This Item

Linear Formula:
C6H4(CH3)2
CAS Number:
Molecular Weight:
106.17
NACRES:
NA.24
UNSPSC Code:
41116107
Beilstein/REAXYS Number:
1815558
MDL number:
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Segment

grade

certified reference material, pharmaceutical secondary standard

agency

traceable to USP 1601849

vapor density

3.7 (vs air)

vapor pressure

<0.1 atm ( 21.1 °C), 16 mmHg ( 37.7 °C), 7 mmHg ( 20 °C)

CofA

current certificate can be downloaded

autoignition temp.

867 °F

expl. lim.

7 %

packaging

pkg of 20 mL

refractive index

n20/D 1.505 (lit.)

bp

143-145 °C (lit.)

mp

-26--23 °C (lit.)

density

0.879 g/mL at 20 °C (lit.)

application(s)

pharmaceutical small molecule

storage temp.

2-30°C

SMILES string

Cc1ccccc1C

InChI

1S/C8H10/c1-7-5-3-4-6-8(7)2/h3-6H,1-2H3

InChI key

CTQNGGLPUBDAKN-UHFFFAOYSA-N

General description

Pharmaceutical secondary standards for application in quality control provide pharma laboratories and manufacturers with a convenient and cost-effective alternative to the preparation of in-house working standards

Analysis Note

These secondary standards offer multi-traceability to the USP and EP primary standards, where they are available.

Other Notes

Find a digital Reference Material for this product available on our online platform ChemisTwin® for NMR. You can use this digital equivalent on ChemisTwin® for your sample identity confirmation and compound quantification (with digital external standard). An NMR spectrum of this substance can be viewed and an online comparison against your sample can be performed with a few mouseclicks. Learn more here and start your free trial.
This Certified Reference Material (CRM) is produced and certified in accordance with ISO 17034 and ISO/IEC 17025. All information regarding the use of this CRM can be found on the certificate of analysis.
To see an example of a Certificate of Analysis for this material enter LRAB9793 in the Documents slot below. This is an example certificate only and may not be the lot that you receive.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
PHR1312X1040XX0020
application(s)

pharmaceutical small molecule

application(s)

pharmaceutical (small molecule)

application(s)

-

application(s)

-

Quality Level

300

Quality Level

300

Quality Level

100

Quality Level

200

grade

certified reference material, pharmaceutical secondary standard

grade

certified reference material, pharmaceutical secondary standard

grade

reagent grade

grade

-

storage temp.

2-30°C

storage temp.

2-30°C

storage temp.

-

storage temp.

room temp

packaging

pkg of 20 mL

packaging

-

packaging

-

packaging

-

mp

-26--23 °C (lit.)

mp

-26--23 °C (lit.)

mp

-26--23 °C (lit.)

mp

-26--23 °C (lit.)


Still not finding the right product?

Explore all of our products under o-Xylene


signalword

Danger

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Aquatic Chronic 3 - Asp. Tox. 1 - Eye Irrit. 2 - Flam. Liq. 3 - Skin Irrit. 2 - STOT SE 3

target_organs

Respiratory system

Storage Class

3 - Flammable liquids

wgk

WGK 2

flash_point_f

87.8 °F

flash_point_c

31 °C



Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters