Skip to Content
Merck

Skip To

RAWP04700

MCE Membrane Filter, 1.2 μm Pore Size

greener alternative

MF-Millipore, filter diam. 47 mm, hydrophilic, white

MCE Membrane Filter, 1.2 μm Pore Size

Synonym(s):

Cellulose nitrate membrane filter discs, Hydrophilic MCE membrane filter cut discs, MF-Millipore Membrane Filter, 1.2 µm pore size, Mixed Cellulose Ester membrane filter discs

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice

About This Item

UNSPSC Code:
40161507
NACRES:
NB.24
eCl@ss:
32031602
Pore size:
1.2 μm pore size, 82 % porosity
Material:
mixed cellulose esters (MCE) membrane, plain filter, white filter
Feature:
hydrophilic
Diam.:
47 mm
Sterility:
non-sterile
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Segment

material

mixed cellulose esters (MCE) membrane, plain filter, white filter

description

47 mm diameter, mixed cellulose esters (MCE) membrane, hydrophilic, white, 100 discs

sterility

non-sterile

feature

hydrophilic

manufacturer/tradename

MF-Millipore, Millipore

sustainability

Greener Alternative Product

parameter

20 L/min-cm2 air flow rate, 270 mL/min-cm2 water flow rate, 75 °C max. temp.

diam.

47 mm

filter diam.

47 mm

thickness

150 μm

gravimetric extractables

5%

color

white

refractive index

n/D 1.512

matrix

MF-Millipore

pore size

1.2 μm pore size, 82 % porosity

bubble point

≥0.76 bar, air with water at 23 °C

greener alternative category

shipped in

ambient

General description

Biologically inert mixtures of cellulose acetate and cellulose nitrate have made MF-Millipore membrane filters one of the most widely used membranes in analytical and research applications.

MF-Millipore filters without Triton surfactant contain minimum amounts of wetting agent and have a lower water extractable content than standard MF-Millipore filters.

Features & Benefits:
- Versatile filter for biological and environmental monitoring applications
- Available in a range of pore sizes, colored black or white, with or without a gridded surface
- Compatible with ethylene oxide, gamma irradiation, and autoclave sterilization methods
Filter Code: RAWP
Need a custom membrane filter? Click here to request a custom membrane filter or unique and large package sizes that may not be available via our website.
We are committed to bringing you Greener Alternative Products, which have been re-engineered to enable more sustainable research solutions. The inner & outer boxes containing the membrane discs are made from bio-circular material allocated using the ISCC mass balance approach. Click here for more information.

Application

Clarification of aqueous solutions

Legal Information

MF-Millipore is a trademark of Merck KGaA, Darmstadt, Germany

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
RAWP01300RAWP03700RAWP02500
material

mixed cellulose esters (MCE) membrane, white filter, plain filter

material

mixed cellulose esters (MCE) membrane, plain filter, white filter

material

mixed cellulose esters (MCE) membrane, plain filter, white filter

material

mixed cellulose esters (MCE) membrane, plain filter, white filter

manufacturer/tradename

MF-Millipore

manufacturer/tradename

MF-Millipore

manufacturer/tradename

MF-Millipore

manufacturer/tradename

MF-Millipore

parameter

20 L/min-cm2 air flow rate, 75 °C max. temp., 270 mL/min-cm2 water flow rate

parameter

20 L/min-cm2 air flow rate, 270 mL/min-cm2 water flow rate, 75 °C max. temp.

parameter

20 L/min-cm2 air flow rate, 270 mL/min-cm2 water flow rate, 75 °C max. temp.

parameter

20 L/min-cm2 air flow rate, 270 mL/min-cm2 water flow rate, 75 °C max. temp.

pore size

1.2 μm pore size, 82 % porosity

pore size

1.2 μm pore size, 82 % porosity

pore size

1.2 μm pore size, 82 % porosity

pore size

1.2 μm pore size, 82 % porosity

sterility

non-sterile

sterility

non-sterile

sterility

-

sterility

non-sterile

feature

hydrophilic

feature

hydrophilic

feature

hydrophilic

feature

hydrophilic


Still not finding the right product?


pictograms

Flame

signalword

Danger

Storage Class

4.1B - Flammable solid hazardous materials

Hazard Classifications

ED ENV 1 - Flam. Sol. 1



Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

Active Filters

    • Review 1
    • Votes 0
    2 out of 5 stars.

    Helpful?

    1. Response from Merck KGaA:

      Thank you for your rating. We encourage customers who experience problems with our products to call or email us for additional technical support. Please visit https://www.sigmaaldrich.com/US/en/support/customer-support to submit a Product Technical Inquiry.