Skip to Content
Merck

Skip To

1.00443

Nitric acid

65%, EMPLURA®

Synonym(s):

Nitric acid solution

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice

About This Item

Empirical Formula (Hill Notation):
HNO3
CAS Number:
Molecular Weight:
63.01
UNSPSC Code:
12352301
NACRES:
NA.21
MDL number:
Concentration:
64.3-66.4% HNO3
Form:
liquid
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Product Name

Nitric acid 65%, EMPLURA®

vapor pressure

9.4 hPa ( 20 °C)

Quality Segment

product line

EMPLURA®

form

liquid

concentration

64.3-66.4% HNO3

impurities

≤0.0001% As (Arsenic), ≤0.0003% Chloride (Cl), ≤0.0004% Fe (Iron), ≤0.0005% Heavy metals (as Pb), ≤0.001% Ca (Calcium), ≤0.001% NH4 (Ammonium), ≤0.001% Sulfate (SO4), ≤0.003% Nitrogen oxides (as N2O3)

pH

<1 (20 °C in H2O, strongly acid)

bp

121 °C/1013 hPa

mp

-32 °C

solubility

water: soluble at 20 °C

density

1.39 g/cm3 at 20 °C

storage temp.

2-30°C

SMILES string

O[N+]([O-])=O

InChI

1S/HNO3/c2-1(3)4/h(H,2,3,4)

InChI key

GRYLNZFGIOXLOG-UHFFFAOYSA-N

Application


  • In-vitro evaluation of microleakage of bioceramic root-end filling materials: A spectrophotometric study.: This research employed 65% nitric acid for sample preparation in the spectrophotometric analysis of microleakage in dental materials, providing insights into the effectiveness of different bioceramic materials (Gupta and Kewalramani, 2021).

  • A comparative evaluation of resin- and varnish-based surface protective agents on glass ionomer cement - a spectrophotometric analysis.: The study used 65% nitric acid for the preparation of glass ionomer cement samples to evaluate the effectiveness of surface protective agents through spectrophotometric methods (Tyagi et al., 2020).

  • Spectrophotometric analysis evaluating apical microleakage in retrograde filling using GIC, MTA and biodentine: an in-vitro study.: This research utilized 65% nitric acid in the preparation of samples for the evaluation of microleakage in dental restorative materials, highlighting the performance of various materials in preventing leakage (Nepal et al., 2020).

  • Influence of stacked structure of carbons modified on its surface on n-pentane adsorption.: This study applied 65% nitric acid for surface modification of carbon materials, investigating the effects on adsorption properties, with significant findings on how surface structure influences adsorption efficiency (Carvajal-Bernal et al., 2019).

Analysis Note

Assay (alkalimetric): 64.3 - 66.4 %
Identity: passes test
Appearance of solution: passes test
Chloride (Cl): ≤ 0.0003 %
Iodate, Bromate: passes test
Nitrogen oxides (as N₂O₃): ≤ 0.003 %
Sulfate (SO₄): ≤ 0.001 %
Heavy metals (as Pb): ≤ 0.0005 %
As (Arsenic): ≤ 0.0001 %
Ca (Calcium): ≤ 0.001 %
Fe (Iron): ≤ 0.0004 %
NH₄ (Ammonium): ≤ 0.001 %
Residual solvents (ICH Q3C): ...excluded by production process
Evaporation residue: ≤ 0.01
Date of expiry: see product label

Legal Information

EMPLURA is a registered trademark of Merck KGaA, Darmstadt, Germany

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
1.004411.151871.01832
concentration

64.3-66.4% HNO3

concentration

65%

concentration

-

concentration

68.0-70.0% HNO3

Quality Level

200

Quality Level

100

Quality Level

100

Quality Level

200

form

liquid

form

liquid

form

liquid

form

liquid

solubility

water: soluble at 20 °C

solubility

water: soluble

solubility

water: soluble at 20 °C

solubility

-

storage temp.

2-30°C

storage temp.

2-30°C

storage temp.

2-30°C

storage temp.

2-30°C

pH

<1 (20 °C in H2O, strongly acid)

pH

<1 (20 °C in H2O, strongly acid)

pH

<1 (20 °C in H2O)

pH

<1 (20 °C in H2O)


Still not finding the right product?

Explore all of our products under Nitric acid 65%


signalword

Danger

Hazard Classifications

Acute Tox. 3 Inhalation - Eye Dam. 1 - Met. Corr. 1 - Ox. Liq. 3 - Skin Corr. 1A

supp_hazards

Storage Class

5.1B - Oxidizing hazardous materials

wgk

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable



Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters