Skip to Content
Merck

Skip To

A-205

Withanone solution

20 μg/mL in methanol, certified reference material, ampule of 1 mL, Cerilliant®

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Pack SizeSKUAvailabilityPrice

About This Item

Empirical Formula (Hill Notation):
C28H38O6
CAS Number:
Molecular Weight:
470.60
UNSPSC Code:
41116107
NACRES:
NA.24
MDL number:
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


grade

certified reference material

Quality Segment

feature

Snap-N-Spike®/Snap-N-Shoot®

packaging

ampule of 1 mL

manufacturer/tradename

Cerilliant®

concentration

20 μg/mL in methanol

format

single component solution

storage temp.

−20°C

SMILES string

O1[C@H](CC(=C(C1=O)C)C)[C@H]([C@@]2([C@@]3([C@H]([C@@H]4[C@@H]5O[C@@H]5[C@@]6([C@@]([C@H]4CC3)(C(=O)C=CC6)C)O)CC2)C)O)C

InChI

1S/C28H38O6/c1-14-13-19(33-24(30)15(14)2)16(3)27(31)12-9-17-21-18(8-11-25(17,27)4)26(5)20(29)7-6-10-28(26,32)23-22(21)34-23/h6-7,16-19,21-23,31-32H,8-13H2,1-5H3/t16-,17+,18+,19-,21+,22+,23+,25+,26+,27+,28+/m1/s1

InChI key

FAZIYUIDUNHZRG-PCTWTJKKSA-N

General description

This phytochemical certified reference materials solution is produced at the Millipore Sigma Cerilliant site in Round Rock Texas under ISO/IEC 17025 and ISO 17034 accreditation and offers a cost efficient and precise tool for calibration and verification of one of the key components for Withania (Ashwagandha), a plant frequently used in traditonal indian medicine (Ayurveda). A comprehensive certificate including traceable values and expanded uncertainties for all the analytes is available.




Discover the entire range of phytochemical certified reference material solutions from Cerilliant

Legal Information

CERILLIANT is a registered trademark of Merck KGaA, Darmstadt, Germany
Snap-N-Shoot is a registered trademark of Cerilliant Corporation
Snap-N-Spike is a registered trademark of Merck KGaA, Darmstadt, Germany

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
1.70244A-1971.19801
grade

certified reference material

grade

certified reference material

grade

certified reference material

grade

certified reference material

format

single component solution

format

single component solution

format

multi-component solution

format

single component solution

feature

Snap-N-Spike®/Snap-N-Shoot®

feature

-

feature

Snap-N-Spike®/Snap-N-Shoot®

feature

-

concentration

20 μg/mL in methanol

concentration

1000 mg/L W in water

concentration

20-100 μg/mL in methanol

concentration

1000 mg/L Tl in 0.5 M HNO3

storage temp.

−20°C

storage temp.

15-25°C

storage temp.

−70°C

storage temp.

15-25°C

manufacturer/tradename

Cerilliant®

manufacturer/tradename

-

manufacturer/tradename

Cerilliant®

manufacturer/tradename

-


signalword

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Flam. Liq. 2 - STOT SE 1

target_organs

Eyes,Central nervous system

Storage Class

3 - Flammable liquids

wgk

WGK 2

flash_point_f

49.5 °F

flash_point_c

9.7 °C



Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Global Trade Item Number

SKUGTIN
R1401-10MG04061833274019
A1487-5MG04061832866451
A1487-25MG04061832866444
33688-1ML04061826743348
AB967604053252333675
Z568600-1EA04061833581230
ERMAE649-5ML04061837904806
Z528749-1EA04061832771298
Z528730-2EA04061832771281
SCC35504065267130972
SCC35404065267124094
SCC35304065267130965
T6694-1VL04061838043757
SAB4504561-100UG04061832526713
769835-5G04061832929712
769835-1G04061826139585
A-205-1ML04065267919706
Z528870-1EA04061832771342
HMI0359-5NMOL04061831392722
HMI0360-5NMOL04061831382150
SAB4200447-200UL04061837875557
C9253-250MG04061833530399
C9253-1G04061833530382

Questions

Reviews

No rating value

Active Filters