Skip to Content
Merck

Skip To

74716

Octaethylene glycol monodecyl ether

BioXtra, ≥98.0% (TLC)

Synonym(s):

C10E8, Decyl octaethylene glycol ether, Decyloctaglycol, Polyoxyethylene (8) decyl ether

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Pack SizeSKUAvailabilityPrice

About This Item

Linear Formula:
CH3(CH2)9(OCH2CH2)8OH
CAS Number:
Molecular Weight:
510.70
UNSPSC Code:
12161900
NACRES:
NA.25
PubChem Substance ID:
MDL number:
Beilstein/REAXYS Number:
4760288
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


description

non-ionic

Quality Level

product line

BioXtra

assay

≥98.0% (TLC)

form

crystals

mol wt

510.70 g/mol

technique(s)

protein quantification: suitable

refractive index

n20/D 1.457

density

1.008 g/mL at 20 °C (lit.)

cation traces

Ca: ≤50 mg/kg, Cd: ≤50 mg/kg, Co: ≤50 mg/kg, Cr: ≤50 mg/kg, Cu: ≤50 mg/kg, Fe: ≤50 mg/kg, K: ≤50 mg/kg, Mg: ≤50 mg/kg, Mn: ≤50 mg/kg, Na: ≤50 mg/kg, Ni: ≤50 mg/kg, Pb: ≤50 mg/kg, Zn: ≤50 mg/kg

SMILES string

CCCCCCCCCCOCCOCCOCCOCCOCCOCCOCCOCCO

InChI

1S/C26H54O9/c1-2-3-4-5-6-7-8-9-11-28-13-15-30-17-19-32-21-23-34-25-26-35-24-22-33-20-18-31-16-14-29-12-10-27/h27H,2-26H2,1H3

InChI key

UJMHIOBAHVUDGS-UHFFFAOYSA-N


Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
747175204476437
technique(s)

protein quantification: suitable

technique(s)

protein purification: suitable

technique(s)

-

technique(s)

drug transporter assay: suitable

form

crystals

form

solid

form

-

form

semisolid

assay

≥98.0% (TLC)

assay

≥98.0% (GC)

assay

≥98.0% (TLC)

assay

≥98.0% (GC)

cation traces

Ca: ≤50 mg/kg, Co: ≤50 mg/kg, Cu: ≤50 mg/kg, K: ≤50 mg/kg, Mn: ≤50 mg/kg, Ni: ≤50 mg/kg, Zn: ≤50 mg/kg, Cr: ≤50 mg/kg, Mg: ≤50 mg/kg, Pb: ≤50 mg/kg, Cd: ≤50 mg/kg, Na: ≤50 mg/kg, Fe: ≤50 mg/kg

cation traces

Ca: ≤50 mg/kg, Cd: ≤50 mg/kg, Co: ≤50 mg/kg, Cr: ≤50 mg/kg, Cu: ≤50 mg/kg, Fe: ≤50 mg/kg, K: ≤50 mg/kg, Mg: ≤50 mg/kg, Mn: ≤50 mg/kg, Na: ≤50 mg/kg, Ni: ≤50 mg/kg, Pb: ≤50 mg/kg, Zn: ≤50 mg/kg

cation traces

Ca: ≤50 mg/kg, Cd: ≤50 mg/kg, Co: ≤50 mg/kg, Cr: ≤50 mg/kg, Cu: ≤50 mg/kg, Fe: ≤50 mg/kg, K: ≤50 mg/kg, Mg: ≤50 mg/kg, Mn: ≤50 mg/kg, Na: ≤50 mg/kg, Ni: ≤50 mg/kg, Pb: ≤50 mg/kg, Zn: ≤50 mg/kg

cation traces

Ca: ≤50 mg/kg, Cd: ≤50 mg/kg, Co: ≤50 mg/kg, Cr: ≤50 mg/kg, Cu: ≤50 mg/kg, Fe: ≤50 mg/kg, K: ≤50 mg/kg, Mg: ≤50 mg/kg, Mn: ≤50 mg/kg, Na: ≤50 mg/kg, Ni: ≤50 mg/kg, Pb: ≤50 mg/kg, Zn: ≤50 mg/kg

density

1.008 g/mL at 20 °C (lit.)

density

-

density

-

density

0.963 g/mL at 20 °C (lit.)

description

non-ionic

description

non-ionic

description

non-ionic

description

non-ionic


Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Soma Das et al.
The Journal of general virology, 98(5), 962-976 (2017-02-22)
Hepatitis C virus (HCV) is a leading cause of chronic viral hepatitis, but an effective vaccine is still not available to prevent infection. Use of neutralizing antibodies could be a potential therapeutic option. In this study, the presence of anti-HCV
Aggeliki Quell et al.
Langmuir : the ACS journal of surfaces and colloids, 33(2), 537-542 (2017-01-07)
During our studies on emulsion-templated monodisperse polymer foams we found significant differences in the finestructure if the locus of initiation is changed. This motivated us to study the phase behavior of the liquid template. Our studies indicate that the template
Sara G Hovakeemian et al.
Soft matter, 11(34), 6840-6851 (2015-08-04)
Most antimicrobial peptides act upon target microorganisms by permeabilizing their membranes. The mode of action is often assessed by vesicle leakage experiments that use model membranes, with the assumption that biological activity correlates with the permeabilization of the lipid bilayer.



Global Trade Item Number

SKUGTIN
74716-1ML04061838169396
74716-5ML04061838169402

Questions

Reviews

No rating value

Active Filters