Select a Size
| Size/SKU | Availability | Price |
|---|
About This Item
grade
pharmaceutical primary standard
agency
USP
API family
levetiracetam
manufacturer/tradename
USP
mp
259-263 °C
application(s)
pharmaceutical (small molecule)
format
neat
SMILES string
CC[C@H](N)C(N)=O.[H]Cl
InChI
1S/C4H10N2O.ClH/c1-2-3(5)4(6)7;/h3H,2,5H2,1H3,(H2,6,7);1H/t3-;/m0./s1
InChI key
HDBMIDJFXOYCGK-DFWYDOINSA-N
General description
Application
- Levetiracetam
- Levetiracetam Tablets
Analysis Note
Other Notes
1 of 1
This Item | |||
|---|---|---|---|
| manufacturer/tradename USP | manufacturer/tradename - | manufacturer/tradename - | manufacturer/tradename USP |
| grade pharmaceutical primary standard | grade certified reference material, pharmaceutical secondary standard | grade certified reference material, pharmaceutical secondary standard | grade pharmaceutical primary standard |
| application(s) pharmaceutical (small molecule) | application(s) pharmaceutical small molecule | application(s) pharmaceutical small molecule | application(s) pharmaceutical (small molecule) |
| format neat | format - | format - | format neat |
| API family levetiracetam | API family levetiracetam | API family levetiracetam | API family levetiracetam |
| mp 259-263 °C | mp 259-263 °C | mp - | mp - |
Still not finding the right product?
Explore all of our products under Levetiracetam Related Compound B
signalword
Warning
hcodes
Hazard Classifications
Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3
target_organs
Respiratory system
Storage Class
11 - Combustible Solids
flash_point_f
Not applicable
flash_point_c
Not applicable
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.



