Skip to Content
Merck

Skip To

G8645

D-Glucuronic acid sodium salt monohydrate

97.5-102.5% (non-aqueous titration)

Synonym(s):

Sodium D-glucuronate

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Pack SizeSKUAvailabilityPrice

About This Item

Empirical Formula (Hill Notation):
C6H9NaO7 · H2O
CAS Number:
Molecular Weight:
234.14
UNSPSC Code:
12352201
NACRES:
NA.25
PubChem Substance ID:
EC Number:
239-065-7
Beilstein/REAXYS Number:
4218577
MDL number:
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


biological source

synthetic (organic

Organic)

Quality Level

assay

97.5-102.5% (non-aqueous titration)

form

powder

color

white to off-white

mp

138 °C (dec.) (lit.)

solubility

water: 50 mg/mL, clear, colorless to light yellow

storage temp.

room temp

SMILES string

O.[Na+].OC1O[C@@H]([C@@H](O)[C@H](O)[C@H]1O)C([O-])=O

InChI

1S/C6H10O7.Na.H2O/c7-1-2(8)4(5(10)11)13-6(12)3(1)9;;/h1-4,6-9,12H,(H,10,11);;1H2/q;+1;/p-1/t1-,2-,3+,4-,6?;;/m0../s1

InChI key

FDXOVWIMYQPHRO-HWZSTGBWSA-M

Biochem/physiol Actions

Glucuronic acid is a carboxylic acid which in involved in the process of glucuronidation, where glucuronic acid is linked to a molecule to allow for more efficient transport due its highly water-soluble nature. Most often it is used to rid the body of toxins or to aid in the delivery of drugs or hormones.

Other Notes

To gain a comprehensive understanding of our extensive range of Monosaccharides for your research, we encourage you to visit our Carbohydrates Category page.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
G9005P7877P6888
assay

97.5-102.5% (non-aqueous titration)

assay

≥99% (HPLC)

assay

≥97% (enzymatic)

assay

≥95% anhydrous basis

biological source

synthetic (organic

Organic)

biological source

microbial

biological source

synthetic (organic)

biological source

synthetic (inorganic)

solubility

water: 50 mg/mL, clear, colorless to light yellow

solubility

590 g/L at 25 °C

solubility

water: 50 mg/mL, clear, colorless

solubility

water: 50 mg/mL, clear, colorless

Quality Level

300

Quality Level

200

Quality Level

200

Quality Level

200

form

powder

form

powder or crystals

form

powder

form

powder

storage temp.

room temp

storage temp.

room temp

storage temp.

−20°C

storage temp.

−20°C


Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Gerrit Schoenicke et al.
Nephrology, dialysis, transplantation : official publication of the European Dialysis and Transplant Association - European Renal Association, 17(11), 1988-1992 (2002-10-29)
The glucose polymer icodextrin has gained a widespread use in peritoneal dialysis especially in patients with low ultrafiltration and high peritoneal transport properties. In patients using a once-daily exchange with icodextrin, a decreased serum amylase activity has been reported. We
Carlos M De Leon-Rodriguez et al.
mSphere, 3(5) (2018-10-26)
Phagosomal acidification is a critical cellular mechanism for the inhibition and killing of ingested microbes by phagocytic cells. The acidic environment activates microbicidal proteins and creates an unfavorable environment for the growth of many microbes. Consequently, numerous pathogenic microbes have



Global Trade Item Number

SKUGTIN
G8645-5G04061833649725
G8645-25G04061832390475
G8645-50G04061832390482
G8645-1KG04061826059647
G8645-100G04061832390451
G8645-10G04061832390468

Questions

Reviews

No rating value

Active Filters