Skip to Content
Merck

Skip To

D4803

Nunc® square bioassay dish

standard height, non-treated surface

Synonym(s):

Nunc® Bioassay Dish, dish petri, dish plastic, petri dishes, Nunc 240835

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Pack SizeSKUAvailabilityPrice

About This Item

NACRES:
NB.22
UNSPSC Code:
41121812
Material:
polystyrene , square
Sterility:
sterile
Binding type:
non-treated surface
Manufacturer/tradename:
Nunc 240835
Packaging:
case of 16 ea
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


material

polystyrene , square

sterility

sterile

feature

lid

packaging

case of 16 ea

manufacturer/tradename

Nunc 240835

L × W × H

245 mm × 245 mm × 25 mm

dish L × W

245 mm × 245 mm

dish capacity

225 mL

binding type

non-treated surface

General description

Available in cell culture-treated and non-treated surfaces, the versatile Nunc® Square BioAssay dishes are useful for screening large numbers of colonies, culturing bacteria and fungi, or providing a contained environment for other culture vessels.
Nunc® Bioassay Dish is useful for screening large numbers of colonies in cloning experiments in molecular biology. It is suitable for culturing bacteria and fungi or providing a contained environment for other culture vessels. Nunc Bioassay Dish can be used for agar diffusion assays.
Suitable for agar diffusion assays. Useful for screening large numbers of colonies in cloning experiments in molecular biology.

Application

  • Drug discovery
  • Toxicology
  • Environmental monitoring
Nunc® Bioassay Dish has been used to determine the total number of colony-forming units of transformed cells. It has also been used in multi-site mutagenesis and comprehensive codon mutagenesis by PFunkel using an ssDNA template.

Features and Benefits

  • External dimensions: 245 x 245 x 25mm
  • Lid height: 9.8mm
  • Optically clear and sterile

Legal Information

Nunc is a registered trademark of Thermo Fisher Scientific or its subsidiaries

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
Z740356Z688819Z688827
sterility

sterile

sterility

sterile

sterility

sterile

sterility

sterile

material

polystyrene , square

material

clear polystyrene

material

polystyrene

material

polystyrene

manufacturer/tradename

Nunc 240835

manufacturer/tradename

Nunc 168381

manufacturer/tradename

Nunc 174902

manufacturer/tradename

Nunc 174905

binding type

non-treated surface

binding type

Nunclon Delta surface

binding type

(UpCell Surface)

binding type

(UpCell Surface)

packaging

case of 16 ea

packaging

pack of 80 ea

packaging

case of 6 ea

packaging

case of 6 ea

L × W × H

245 mm × 245 mm × 25 mm

L × W × H

-

L × W × H

-

L × W × H

-


Still not finding the right product?

Explore all of our products under Nunc® square bioassay dish




Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Global Trade Item Number

SKUGTIN
D4803-1CS04061826744406