Skip to Content
Merck

Skip To

ABE26

Anti-acetyl-c-Myc (Lys323) Antibody

from rabbit, purified by affinity chromatography

Synonym(s):

Myc proto-oncogene protein, Class E basic helix-loop-helix protein 39, HLHe39, Proto-oncogene c-Myc, Transcription factor p64, c-MYC

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Pack SizeSKUAvailabilityPrice

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41
eCl@ss:
32160702
Clone:
polyclonal
Species reactivity:
mouse, human
Application:
WB
Citations:
6
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


biological source

rabbit

Quality Level

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

purified by

affinity chromatography

species reactivity

mouse, human

species reactivity (predicted by homology)

rhesus macaque (based on 100% sequence homology), chimpanzee (based on 100% sequence homology)

technique(s)

western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

acetylation (Lys323)

Gene Information

chimpanzee ... Myc(464393)
human ... MYC(4609)
mouse ... Myc(17869)
rhesus monkey ... Myc(694626)

General description

c-Myc (bHLHe39) is a transcription factor that regulates the expression of multiple genes involved in apoptosis, cell growth, differentiation, and proliferation. Active c-Myc is coupled to the MAX protein via c-Myc’s C-terminal helix-loop-helix leucine zipper (bHLHLZ) domain. The c-Myc-MAX heterodimer also interacts with a number of other transcription-related proteins including TRRAP, p107 and Miz-1, and c-Myc-MAX may indirectly regulate histone acetylases involved in chromatin remodeling. c-Myc is itself acetylated on lysine 323 by GCN5, and on lysine 148 and 157 by p300/CBP. The significance of c-Myc acetylation is still under investigation; however, previous studies have suggested that HAT-dependent acetylation of c-Myc may regulate its turnover and stability. c-Myc may play a role in various cancers such as Burkitt’s lymphoma.
~58 kDa observed. Uniprot describes two isoforms at 49 and 51 kDa

Immunogen

Epitope: Acetylated Lys323
KLH-conjugated linear peptide corresponding to human c-Myc acetylated at Lys323.

Application

Peptide Inhibition Assay Analysis: 2 µg/mL from a representative lot blocked acetyl-c-Myc (Lys323) in HEK293T co-transfected with wild type c-Myc or mutant c-Myc (Lys323R) and P300.

Peptide Inhibition Assay Analysis: A representative lot was used by an independent laboratory in HEK293T cells co-transfected with mouse wildtype c-Myc and P300. (Image courtesy of Dr. Ernest Martinez, Department of Biochemistry, University of California at Riverside.)
Research Category
Epigenetics & Nuclear Function
Research Sub Category
Transcription Factors
Use Anti-acetyl-c-Myc (Lys323) Antibody (Rabbit Polyclonal Antibody) validated in WB to detect acetyl-c-Myc (Lys323) also known as HLHe39, Proto-oncogene c-Myc, Transcription factor p64, c-MYC.

Biochem/physiol Actions

Other homologies: Rat, Feline, Sheep, and Bovine (90% sequence homology).
This antibody recognizes c-Myc acetylated at Lys323.

Physical form

Affinity purified
Purified rabbit polyclonal in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Preparation Note

Stable for 1 year at 2-8°C from date of receipt.

Analysis Note

Control
Transfected HEK293T cell lysates
Evaluated by Western Blot in transfected HEK293T cell lysates.

Western Blot Analysis: 0.2 µg/mL of this antibody detected c-Myc on 10 µg of transfected HEK293T cell lysates.

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.


Still not finding the right product?

Try our Product Selector Tool to narrow your options


Storage Class

12 - Non Combustible Liquids

wgk

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable



Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library


Related Content

New Products: Antibodies, Assays, Small Molecules, Inhibitors, and Proteins


Matthew Hurd et al.
Genes & development, 37(19-20), 865-882 (2023-10-19)
The MYC oncogenic transcription factor is acetylated by the p300 and GCN5 histone acetyltransferases. The significance of MYC acetylation and the functions of specific acetylated lysine (AcK) residues have remained unclear. Here, we show that the major p300-acetylated K148(149) and
Wesley W Wang et al.
Journal of the American Chemical Society, 143(40), 16700-16708 (2021-10-01)
Protein acetylation is a central event in orchestrating diverse cellular processes. However, current strategies to investigate protein acetylation in cells are often nonspecific or lack temporal and magnitude control. Here, we developed an acetylation tagging system, AceTAG, to induce acetylation
David Ngai et al.
Nature metabolism, 5(12), 2206-2219 (2023-11-28)
The clearance of apoptotic cells by macrophages (efferocytosis) prevents necrosis and inflammation and activates pro-resolving pathways, including continual efferocytosis. A key resolution process in vivo is efferocytosis-induced macrophage proliferation (EIMP), in which apoptotic cell-derived nucleotides trigger Myc-mediated proliferation of pro-resolving



Global Trade Item Number

SKUGTIN
ABE2604053252522178

Questions

Reviews

No rating value

Active Filters