Skip to Content
MilliporeSigma

Skip To

1623411

USP

Succinic acid

United States Pharmacopeia (USP) Reference Standard

Synonym(s):

Butanedioic acid

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Pack SizeSKUAvailabilityPrice

About This Item

Linear Formula:
HOOCCH2CH2COOH
CAS Number:
Molecular Weight:
118.09
UNSPSC Code:
41116107
NACRES:
NA.24
PubChem Substance ID:
MDL number:
Beilstein/REAXYS Number:
1754069
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


grade

pharmaceutical primary standard

API family

succinic acid

form

crystalline

manufacturer/tradename

USP

bp

235 °C (lit.)

mp

184-186 °C (lit.)

solubility

water: soluble 83 g/L at 25 °C

density

1.564 g/cm3 at 15 °C

application(s)

pharmaceutical (small molecule)

format

neat

SMILES string

OC(=O)CCC(O)=O

InChI

1S/C4H6O4/c5-3(6)1-2-4(7)8/h1-2H2,(H,5,6)(H,7,8)

InChI key

KDYFGRWQOYBRFD-UHFFFAOYSA-N

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia.For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Succinic acid USP reference standard suitable for use in specified USP compendial quality tests and assays.

Also used to prepare standard, and system suitability solution for the assay of succinic acid by using liquid chromatography coupled with UV detector.

Analysis Note

These products are for test and assay use only. They are not meant for administration to humans or animals and cannot be used to diagnose, treat, or cure diseases of any kind.  ​

Other Notes

Sales restrictions may apply.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
49893S7501W471912
grade

pharmaceutical primary standard

grade

certified reference material, TraceCERT®

grade

-

grade

FG, Halal, Kosher, natural

manufacturer/tradename

USP

manufacturer/tradename

Manufactured by: Sigma-Aldrich Production GmbH, Switzerland

manufacturer/tradename

-

manufacturer/tradename

-

application(s)

pharmaceutical (small molecule)

application(s)

cleaning products
cosmetics
flavors and fragrances
food and beverages
personal care

application(s)

-

application(s)

flavors and fragrances

format

neat

format

neat

format

-

format

-

form

crystalline

form

-

form

-

form

crystals

solubility

water: soluble 83 g/L at 25 °C

solubility

-

solubility

H2O: soluble 1 g/10 mL

solubility

water: 83 g/L at 25 °C


Still not finding the right product?

Explore all of our products under Succinic acid


pictograms

Corrosion

signalword

Danger

hcodes

Hazard Classifications

Eye Dam. 1

Storage Class

11 - Combustible Solids

wgk

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Succinic Acid
USP43-NF38: United States Pharmacopeia and National Formulary
United States Pharmacopeia, 39(2), 6074-6074 (2020)



Global Trade Item Number

SKUGTIN
162341104061833847169
1623411-100MG04061838746924
1623411-300MG04061838670342

Questions

Reviews

No rating value

Active Filters