Skip to Content
MilliporeSigma

Skip To

1614501

USP

Sodium nitroprusside

United States Pharmacopeia (USP) Reference Standard

Synonym(s):

Sodium nitroferricyanide(III) dihydrate, Nitroprusside sodium, SNP, Sodium nitroprusside, Sodium pentacyanonitrosylferrate

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Pack SizeSKUAvailabilityPrice

About This Item

Linear Formula:
Na2[Fe(CN)5NO] · 2H2O
CAS Number:
Molecular Weight:
297.95
NACRES:
NA.24
PubChem Substance ID:
UNSPSC Code:
41116107
MDL number:
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


grade

pharmaceutical primary standard

API family

sodium nitroprusside

manufacturer/tradename

USP

application(s)

pharmaceutical (small molecule)

format

neat

storage temp.

2-8°C

SMILES string

[Na+].[Na+].[H]O[H].[H]O[H].O=N[Fe--](C#N)(C#N)(C#N)(C#N)C#N

InChI

1S/5CN.Fe.NO.2Na.2H2O/c5*1-2;;1-2;;;;/h;;;;;;;;;2*1H2/q;;;;;2*-1;2*+1;;

InChI key

OIRZWVYIQXBRFC-UHFFFAOYSA-N

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia.For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Sodium nitroprusside USP reference standard, intended for use in specified quality tests and assays as specified in the USP compendia.

Also, for use with USP monograph such as Sodium Nitroprusside for Injection

Analysis Note

These products are for test and assay use only. They are not meant for administration to humans or animals and cannot be used to diagnose, treat, or cure diseases of any kind.  ​

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
PHR142371778567538
grade

pharmaceutical primary standard

grade

certified reference material, pharmaceutical secondary standard

grade

ACS reagent, puriss. p.a.

grade

-

manufacturer/tradename

USP

manufacturer/tradename

-

manufacturer/tradename

-

manufacturer/tradename

Calbiochem®

application(s)

pharmaceutical (small molecule)

application(s)

pharmaceutical (small molecule)

application(s)

-

application(s)

-

format

neat

format

neat

format

-

format

-

storage temp.

2-8°C

storage temp.

2-8°C

storage temp.

-

storage temp.

10-30°C

API family

sodium nitroprusside

API family

sodium nitroprusside

API family

-

API family

-


Still not finding the right product?

Explore all of our products under Sodium nitroprusside


pictograms

Skull and crossbones

signalword

Danger

hcodes

Hazard Classifications

Acute Tox. 3 Oral

Storage Class

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Xiao Guan et al.
Acta pharmacologica Sinica (2021-10-02)
The multi-generation heredity trait of hypertension in human has been reported, but the molecular mechanisms underlying multi-generational inheritance of hypertension remain obscure. Recent evidence shows that prenatal inflammatory exposure (PIE) results in increased incidence of cardiovascular diseases, including hypertension. In
Christina E Larder et al.
Nutrients, 13(8) (2021-08-28)
Osteoarthritis (OA), the most common form of arthritis, is associated with metabolic diseases and gut microbiome dysbiosis. OA patients often take supplements of collagen hydrolysates (CHs) with a high peptide content. Following digestion, some peptides escape absorption to induce prebiotic



Global Trade Item Number

SKUGTIN
1614501-500MG04061838746344

Questions

Reviews

No rating value

Active Filters