Skip to Content
MilliporeSigma

Skip To

1478877

USP

ORLISTAT RELATED COMPOUND D SOLUTION

United States Pharmacopeia (USP) Reference Standard

Synonym(s):

N-Formyl-L-leucine (3S,4R,6S)-3-hexyl-2-oxo-6-undecyltetrahydro-2H-pyran-4-yl ester

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Pack SizeSKUAvailabilityPrice

About This Item

Empirical Formula (Hill Notation):
C29H53NO5
CAS Number:
Molecular Weight:
495.73
MDL number:
UNSPSC Code:
41116107
NACRES:
NA.24
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


grade

pharmaceutical primary standard

agency

USP

API family

orlistat

manufacturer/tradename

USP

application(s)

pharmaceutical

format

neat

SMILES string

N([C@@H](CC(C)C)C(=O)O[C@H]1[C@@H](C(=O)O[C@H](C1)CCCCCCCCCCC)CCCCCC)C=O

InChI

1S/C29H53NO5/c1-5-7-9-11-12-13-14-15-16-18-24-21-27(25(28(32)34-24)19-17-10-8-6-2)35-29(33)26(30-22-31)20-23(3)4/h22-27H,5-21H2,1-4H3,(H,30,31)/t24-,25-,26-,27+/m0/s1

InChI key

IYWNIYFHQZQPBP-YIPNQBBMSA-N

General description

This Certified Reference Material (CRM) is produced and certified in accordance with ISO 17034 and ISO/IEC 17025. All information regarding the use of this CRM can be found on the certificate of analysis.

Application

Orlistat Related Compound Solution USP Reference standard, intended for use in specified quality tests and assays as specified in the USP compendia. Also, for use with USP monographs such as:
  • Orlistat
  • Orlistat Capsules

Analysis Note

These products are for test and assay use only. They are not meant for administration to humans or animals and cannot be used to diagnose, treat, or cure diseases of any kind.

Other Notes

Sales restrictions may apply.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
161588711632641604563
grade

pharmaceutical primary standard

grade

-

grade

-

grade

-

manufacturer/tradename

USP

manufacturer/tradename

USP

manufacturer/tradename

USP

manufacturer/tradename

USP

application(s)

pharmaceutical

application(s)

pharmaceutical (small molecule)

application(s)

pharmaceutical (small molecule)

application(s)

pharmaceutical (small molecule)

format

neat

format

neat

format

neat

format

neat

API family

orlistat

API family

sorafenib

API family

dapagliflozin

API family

rivaroxaban

agency

USP

agency

-

agency

-

agency

-


pictograms

FlameExclamation mark

signalword

Danger

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Eye Irrit. 2 - Flam. Liq. 2

Storage Class

3 - Flammable liquids

flash_point_f

35.6 °F - closed cup

flash_point_c

2.0 °C - closed cup



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Global Trade Item Number

SKUGTIN
1478877-2X1ML04065268894156

Questions

Reviews

No rating value

Active Filters