Skip to Content
MilliporeSigma

Skip To

1445547

USP

USP mAb 002, Monoclonal IgG1

United States Pharmacopeia (USP) Reference Standard

Synonym(s):

Monoclonal IgG1

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Pack SizeSKUAvailabilityPrice

About This Item

CAS Number:
MDL number:
NACRES:
NA.24
UNSPSC Code:
41116107
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


grade

pharmaceutical primary standard

agency

USP

form

liquid

mol wt

~150,000 Da

packaging

pkg of 2 mg

manufacturer/tradename

USP

application(s)

USP Biologics
pharmaceutical

format

neat

storage temp.

−70°C

General description

USP mAb 002 is a second recombinant IgG1 monoclonal antibody standard provided by USP. It is used to evaluate analytical method robustness, control critical quality attributes (CQAs), and compare biosimilar candidates against originator monoclonal antibodies.

The USP biologics monoclonal antibodies (mAb) category includes a range of therapeutic proteins that are designed to target specific antigens, providing effective treatments for various diseases, including cancers and autoimmune disorders. Monoclonal antibodies are produced using hybridoma technology and are essential in both diagnostics and therapeutics. The USP provides comprehensive standards, reference materials, and analytical procedures to ensure the identity, quality, purity, and consistency of monoclonal antibody products throughout their lifecycle.

The United States Pharmacopeia (USP) provides quality standards for biologics to ensure their safety, efficacy, and quality throughout the manufacturing process. These standards assist manufacturers in adhering to regulatory requirements and help safeguard public health by reducing risks associated with biologics.

Application

  • Size-Exclusion Chromatography
  • Capillary SDS Electrophoresis (Reduced and Nonreduced)
  • Analysis of N-Linked Oligosaccharides
  • Sialic Acid Analysis
  • Intact Mass
  • Sequence identification by peptide mapping
  • Independent control material for method development, training, and method transfer
  • Internal assay control for standardization of physico-chemical testing

Features and Benefits

  • 4 Unique proteins with unique quality attributes
  • Extensively characterized for confidence in method development
  • Technical Note available

Analysis Note

These products are for test and assay use only. They are not meant for administration to humans or animals and cannot be used to diagnose, treat, or cure diseases of any kind.  

Other Notes

Sales restrictions may apply.
These products underwent USP′s rigorous collaborative testing process that including assessment by the USP General Chapter 129 methods.
This product is part of the USP Biologics program.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
14455391445595SAB4200683
manufacturer/tradename

USP

manufacturer/tradename

USP

manufacturer/tradename

USP

manufacturer/tradename

-

format

neat

format

neat

format

neat

format

-

application(s)

USP Biologics
pharmaceutical

application(s)

USP Biologics
pharmaceutical

application(s)

USP Biologics
pharmaceutical

application(s)

-

form

liquid

form

liquid

form

liquid

form

buffered aqueous solution

storage temp.

−70°C

storage temp.

−70°C

storage temp.

−70°C

storage temp.

−20°C

packaging

pkg of 2 mg

packaging

pkg of 2 mg

packaging

pkg of 2 mg

packaging

-


Storage Class

12 - Non Combustible Liquids

wgk

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Global Trade Item Number

SKUGTIN
1445547-2MG04065267682662

Questions

Reviews

No rating value

Active Filters