Skip to Content
MilliporeSigma

Skip To

1329800

USP

Hydroxypropyl cellulose

United States Pharmacopeia (USP) Reference Standard

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Pack SizeSKUAvailabilityPrice

About This Item

CAS Number:
UNSPSC Code:
41116107
NACRES:
NA.24
MDL number:
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


grade

pharmaceutical primary standard

API family

hydroxypropyl cellulose

autoignition temp.

752 °F

manufacturer/tradename

USP

density

0.5 g/mL at 25 °C (lit.)

application(s)

pharmaceutical (small molecule)

format

neat

SMILES string

N(C)(C)c1cc(c(cc1)C(N)CC)C

InChI

1S/C12H20N2/c1-5-12(13)11-7-6-10(14(3)4)8-9(11)2/h6-8,12H,5,13H2,1-4H3

InChI key

RRHXDYJWVYFMKV-UHFFFAOYSA-N

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia.For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Hydroxypropyl cellulose USP reference standard, intended for use in specified quality tests and assays as specified in the USP compendia. Also, for use with USP monographs such as:
  • Hydroxypropyl Cellulose Ocular System

Analysis Note

These products are for test and assay use only. They are not meant for administration to humans or animals and cannot be used to diagnose, treat, or cure diseases of any kind.  ​

Other Notes

Sales restrictions may apply.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
1329822191906Y0001720
manufacturer/tradename

USP

manufacturer/tradename

USP

manufacturer/tradename

-

manufacturer/tradename

EDQM

grade

pharmaceutical primary standard

grade

pharmaceutical primary standard

grade

-

grade

pharmaceutical primary standard

application(s)

pharmaceutical (small molecule)

application(s)

pharmaceutical

application(s)

-

application(s)

pharmaceutical (small molecule)

format

neat

format

neat

format

-

format

neat

autoignition temp.

752 °F

autoignition temp.

752 °F

autoignition temp.

752 °F

autoignition temp.

752 °F

API family

hydroxypropyl cellulose

API family

-

API family

-

API family

hydroxypropyl cellulose


Still not finding the right product?

Explore all of our products under Hydroxypropyl cellulose


Storage Class

11 - Combustible Solids

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Arden H Wander et al.
The ocular surface, 7(3), 154-162 (2009-07-29)
Options for extending the duration of tear film protection in dry eyes include artificial tear formulations with enhanced viscosity/polymeric systems, ocular ointments and gels, and, recently, the hydroxypropyl cellulose ophthalmic insert (Lacrisert(R); distributed by Aton Pharma, Inc., Lawrenceville, NJ, USA).



Global Trade Item Number

SKUGTIN
1329800-500MG04061838729262

Questions

Reviews

No rating value

Active Filters