Skip to Content
MilliporeSigma

Skip To

1162308

USP

Dacarbazine

United States Pharmacopeia (USP) Reference Standard

Synonym(s):

5-(3,3-Dimethyl-1-triazenyl)imidazole-4-carboxamide, DTIC

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Pack SizeSKUAvailabilityPrice

About This Item

Empirical Formula (Hill Notation):
C6H10N6O
CAS Number:
Molecular Weight:
182.18
NACRES:
NA.24
PubChem Substance ID:
UNSPSC Code:
41116107
MDL number:
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


grade

pharmaceutical primary standard

API family

dacarbazine

manufacturer/tradename

USP

application(s)

pharmaceutical (small molecule)

format

neat

storage temp.

2-8°C

SMILES string

CN(C)\N=N\c1[nH]cnc1C(N)=O

InChI

1S/C6H10N6O/c1-12(2)11-10-6-4(5(7)13)8-3-9-6/h3H,1-2H3,(H2,7,13)(H,8,9)/b11-10+

InChI key

FDKXTQMXEQVLRF-ZHACJKMWSA-N

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia.
For further information and support please go to the website of the issuing Pharmacopoeia.

Analysis Note

These products are for test and assay use only. They are not meant for administration to humans or animals and cannot be used to diagnose, treat, or cure diseases of any kind.  ​

Other Notes

Sales restrictions may apply.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
BP100Y0000733PHR1924
grade

pharmaceutical primary standard

grade

pharmaceutical primary standard

grade

pharmaceutical primary standard

grade

certified reference material, pharmaceutical secondary standard

manufacturer/tradename

USP

manufacturer/tradename

BP

manufacturer/tradename

EDQM

manufacturer/tradename

-

application(s)

pharmaceutical (small molecule)

application(s)

pharmaceutical
pharmaceutical small molecule

application(s)

pharmaceutical (small molecule)

application(s)

pharmaceutical

format

neat

format

neat

format

neat

format

-

API family

dacarbazine

API family

dacarbazine

API family

dacarbazine

API family

dacarbazine

storage temp.

2-8°C

storage temp.

2-8°C

storage temp.

2-8°C

storage temp.

2-8°C


Still not finding the right product?

Explore all of our products under Dacarbazine


signalword

Danger

Storage Class

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

flash_point_f

Not applicable

flash_point_c

Not applicable

pictograms

Health hazard

hcodes

Hazard Classifications

Carc. 1B - Muta. 1B



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Global Trade Item Number

SKUGTIN
1162308-125MG04061833842799

Questions

Reviews

No rating value

Active Filters